Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ASAP2 Antibodies

          Abnova

          DDEF2 Polyclonal Antibody, MaxPab™

          View all (9) ASAP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite DDEF2 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          DDEF2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DDEF2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DDEF2 Antibody (H00008853-B01P) in WB

          Western Blot analysis of ASAP2 expression in transfected 293T cell line by ASAP2 MaxPab polyclonal antibody. Lane1:DDEF2 transfected lysate(110.66 KDa). Lane2:Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DDEF2 Antibody in Western Blot (WB)

          Product Details

          H00008853-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          DDEF2 (NP_003878.1, 1 a.a. approximately 1006 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MPDQISVSEF VAETHEDYKA PTASSFTTRT AQCRNTVAAI EEALDVDRMV LYKMKKSVKA INSSGLAHVE NEEQYTQALE KFGGNCVCRD DPDLGSAFLK FSVFTKELTA LFKNLIQNMN NIISFPLDSL LKGDLKGVKG DLKKPFDKAW KDYETKITKI EKEKKEHAKL HGMIRTEISG AEIAEEMEKE RRFFQLQMCE YLLKVNEIKI KKGVDLLQNL IKYFHAQCNF FQDGLKAVES LKPSIETLST DLHTIKQAQD EERRQLIQLR DILKSALQVE QKEDSQIRQS TAYSLHQPQG NKEHGTERNG SLYKKSDGIR KVWQKRKCSV KNGFLTISHG TANRPPAKLN LLTCQVKTNP EEKKCFDLIS HDRTYHFQAE DEQECQIWMS VLQNSKEEAL NNAFKGDDNT GENNIVQELT KEIISEVQRM TGNDVCCDCG APDPTWLSTN LGILTCIECS GIHRELGVHY SRMQSLTLDV LGTSELLLAK NIGNAGFNEI MECCLPAEDS VKPNPGSDMN ARKDYITAKY IERRYARKKH ADNAAKLHSL CEAVKTRDIF GLLQAYADGV DLTEKIPLAN GHEPDETALH LAVRSVDRTS LHIVDFLVQN SGNLDKQTGK GSTALHYCCL TDNAECLKLL LRGKASIEIA NESGETPLDI AKRLKHEHCE ELLTQALSGR FNSHVHVEYE WRLLHEDLDE SDDDMDEKLQ PSPNRREDRP ISFYQLGSNQ LQSNAVSLAR DAANLAKEKQ RAFMPSILQN ETYGALLSGS PPPAQPAAPS TTSAPPLPPR NVGKVQTASS ANTLWKTNSV SVDGGSRQRS SSDPPAVHPP LPPLRVTSTN PLTPTPPPPV AKTPSVMEAL SQPSKPAPPG ISQIRPPPLP PQPPSRLPQK KPAPGADKST PLTNKGQPRG PVDLSATEAL GPLSNAMVLQ PPAPMPRKSQ ATKLKPKRVK ALYNCVADNP DELTFSEGDV IIVDGEEDQE WWIGHIDGDP GRKGAFPVSF VHFIAD

          Target Information

          ASAP2 is a multidomain protein belonging to the beta family with two ANK repeats, an Arf-GAP domain, a PH domain and a SH3 domain. This protein localizes in the Golgi apparatus and also at the plasma membrane where it colocalizes with protein tyrosine kinase 2-beta (PYK2) and forms a stable complex with PYK2 in vivo. ASAP2 is tyrosine phosphorylated by activated PYK2. It functions as a substrate and downstream target for PYK2 and SRC, a pathway that may be involved in the regulation of vesicular transport. ASAP2 activates the small GTPases ARF1, ARF5 and ARF6 and regulates the formation of post-Golgi vesicles and modulates constitutive secretion. It modulates phagocytosis mediated by Fc gamma receptor and ARF6. Usually detected in heart, brain, placenta, kidney, monocytes and pancreas, it is up-regulated during monocyte maturation. Multiple transcript variants encoding different isoforms have been found.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 2; centaurin, beta 3; development and differentiation enhancing factor 2; Development and differentiation-enhancing factor 2; PAG3; PAP; PAPbeta; Paxillin-associated protein with ARF GAP activity 3; PYK2 C terminus-associated protein; Pyk2 C-terminus-associated protein

          View more View less

          Gene Aliases: AMAP2; ASAP2; CENTB3; DDEF2; KIAA0400; PAG3; PAP; Pap-alpha; SHAG1

          View more View less

          UniProt ID: (Human) O43150

          View more View less

          Entrez Gene ID: (Human) 8853

          View more View less

          Function(s)
          GTPase activator activity protein binding enzyme activator activity metal ion binding
          Process(es)
          positive regulation of GTPase activity
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0