Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • DDAH2 Antibodies

          Proteintech

          DDAH2 Monoclonal Antibody (1A10G3)

          View all (17) DDAH2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite DDAH2 Monoclonal Antibody (1A10G3)

          • Antibody Testing Data (3)
          DDAH2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DDAH2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DDAH2 Antibody (68154-1-IG) in WB

          Various lysates were subjected to SDS PAGE followed by western blot with 68154-1-Ig (DDAH2 antibody) at dilution of 1:10000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DDAH2 Antibody in Western Blot (WB)
          DDAH2 Antibody in Western Blot (WB)
          DDAH2 Antibody in Western Blot (WB)

          Product Details

          68154-1-IG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000-1:50,000
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rabbit, Rat

          Host/Isotype

          Mouse / IgG3

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1A10G3

          Immunogen

          Recombinant protein
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Protein A

          Storage buffer

          PBS with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: MGTPGEGLGR CSHALIRGVP ESLASGEGAG AGLPALDLAK AQREHGVLGG KLRQRLGLQL LELPPEESLP LGPLLGDTAV IQGDTALITR PWSPARRPEV DGVRKALQDL GLRIVEIGDE NATLDGTDVL FTGREFFVGL SKWTNHRGAE IVADTFRDFA VSTVPVSGPS HLRGLCGMGG PRTVVAGSSD AAQKAVRAMA VLTDHPYASL TLPDDAAADC LFLRPGLPGV PPFLLHRGGG DLPNSQEALQ KLSDVTLVPV SCSELEKAGA GLSSLCLVLS TRPHS

          Target Information

          This gene belongs to the dimethylarginine dimethylaminohydrolase (DDAH) gene family. The encoded enzyme plays a role in nitric oxide generation by regulating cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Clone 7u; DADB-110M10.5; DDAH-2; DDAHII; dimethylargininase-2; dimethylarginine dimethylaminohydrolase 2; dimethylarginine dimethylaminohydrolase II; epididymis secretory protein Li 277; Inactive N(G),N(G)-dimethylarginine dimethylaminohydrolase 2; N(G),N(G)-dimethylarginine dimethylaminohydrolase 2; OTTHUMP00000029307; OTTHUMP00000062666; OTTHUMP00000174406; Protein G6a; Putative hydrolase DDAH2; S-phase protein; testis tissue sperm-binding protein Li 54e

          View more View less

          Gene Aliases: 1110003M04Rik; AU019324; AW413173; DDAH; DDAH-2; DDAH2; DDAHII; G6A; HEL-S-277; NG30

          View more View less

          UniProt ID: (Human) O95865, (Mouse) Q99LD8, (Rat) Q6MG60

          View more View less

          Entrez Gene ID: (Human) 23564, (Mouse) 51793, (Rat) 294239, (Rabbit) 100352259

          View more View less

          Function(s)
          catalytic activity protein binding dimethylargininase activity amino acid binding hydrolase activity hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in linear amidines
          Process(es)
          citrulline metabolic process arginine metabolic process arginine catabolic process nitric oxide biosynthetic process nitric oxide mediated signal transduction negative regulation of apoptotic process positive regulation of nitric oxide biosynthetic process regulation of nitric-oxide synthase activity
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-588f58b695-w7k2p:80/100.66.134.75:80.
          git-commit: b8205085541c8e5054bc4cc0f27ee9f88e66ec64
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0