Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Contactin 2 Antibodies

          Invitrogen

          Contactin 2 Recombinant Rabbit Monoclonal Antibody (4V7O8)

          View all (19) Contactin 2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Contactin 2 Recombinant Rabbit Monoclonal Antibody (4V7O8)

          • Antibody Testing Data (1)
          Contactin 2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Contactin 2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Contactin 2 Antibody (MA5-55420) in WB

          Western Blot analysis of various lysates using Contactin 2 Recombinant Monoclonal Antibody (Product # MA5-55420) at 1:20,000 dilution. Secondary antibody: HRP-conjugated Goat anti-Rabbit IgG (H+L) at 1:10,000 dilution. Lysates/proteins: 25 μg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Enhanced Kit. Exposure time: 180s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Contactin 2 Antibody in Western Blot (WB)

          Product Details

          MA5-55420

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000-1:20,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          4V7O8

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 31-1012 of human Contactin 2.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 0.05% BSA, 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          RRID

          AB_3667661

          Product Specific Information

          Immunogen Sequence: SQTTFGPVFE DQPLSVLFPE ESTEEQVLLA CRARASPPAT YRWKMNGTEM KLEPGSRHQL VGGNLVIMNP TKAQDAGVYQ CLASNPVGTV VSREAILRFG FLQEFSKEER DPVKAHEGWG VMLPCNPPAH YPGLSYRWLL NEFPNFIPTD GRHFVSQTTG NLYIARTNAS DLGNYSCLAT SHMDFSTKSV FSKFAQLNLA AEDTRLFAPS IKARFPAETY ALVGQQVTLE CFAFGNPVPR IKWRKVDGSL SPQWTTAEPT LQIPSVSFED EGTYECEAEN SKGRDTVQGR IIVQAQPEWL KVISDTEADI GSNLRWGCAA AGKPRPTVRW LRNGEPLASQ NRVEVLAGDL RFSKLSLEDS GMYQCVAENK HGTIYASAEL AVQALAPDFR LNPVRRLIPA ARGGEILIPC QPRAAPKAVV LWSKGTEILV NSSRVTVTPD GTLIIRNISR SDEGKYTCFA ENFMGKANST GILSVRDATK ITLAPSSADI NLGDNLTLQC HASHDPTMDL TFTWTLDDFP IDFDKPGGHY RRTNVKETIG DLTILNAQLR HGGKYTCMAQ TVVDSASKEA TVLVRGPPGP PGGVVVRDIG DTTIQLSWSR GFDNHSPIAK YTLQARTPPA GKWKQVRTNP ANIEGNAETA QVLGLTPWMD YEFRVIASNI LGTGEPSGPS SKIRTREAAP SVAPSGLSGG GGAPGELIVN WTPMSREYQN GDGFGYLLSF RRQGSTHWQT ARVPGADAQY FVYSNESVRP YTPFEVKIRS YNRRGDGPES LTALVYSAEE EPRVAPTKVW AKGVSSSEMN VTWEPVQQDM NGILLGYEIR YWKAGDKEAA ADRVRTAGLD TSARVSGLHP NTKYHVTVRA YNRAGTGPAS PSANATTMKP PPRRPPGNIS WTFSSSSLSI KWDPVVPFRN ESAVTGYKML YQNDLHLTPT LHLTGKNWIE IPVPEDIGHA LVQIRTTGPG GDGIPAEVHI VRNGGTSMMV EN

          Target Information

          Contactin 2 (CNTN2, transiently-expressed axonal glycoprotein, AXT, TAX, TAX1, TAG-1, axonin 1) is a neuronal cell adhesion molecule (CAM) that influences the formation of axon connections in the developing nervous system. Contactin 2 is a member of the immunoglobulin superfamily (IgSF) and contains a glycosylphosphatidylinositol-anchor, six immunoglobulin (Ig)-like and four Fibronectin type III (FNIII)-like domains. Contactin 2 is expressed predominantly during neural development on the cell membrane of axons in nerve fiber tracts in order to guide commissural axons without promoting their growth. Contactin 2 binds with NgCAM in the plane of the same membrane (cis-binding). The Contactin 2 heterophilic (Contactin 2/NgCAM and Contactin 2/NrCAM) binding sites are localized to the first four Ig domains. The Contactin 2 homophilic (Contactin 2/Contactin 2) binding site is localized to the FNIII domain.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Axonal glycoprotein TAG-1; axonin; Axonin 1; Axonin-1; axonin-1 cell adhesion molecule; Axonin1; contactin 2 (axonal); contactin 2 (transiently expressed); Contactin-2; Contactin2; DKFZp781D102; MGC157722; TAX-1; Transient axonal glycoprotein 1; transiently-expressed axonal glycoprotein

          View more View less

          Gene Aliases: AXT; CNTN2; D130012K04Rik; FAME5; TAG-1; TAG-564; TAG1; TAX; TAX1

          View more View less

          UniProt ID: (Human) Q02246, (Mouse) Q61330, (Rat) P22063

          View more View less

          Entrez Gene ID: (Human) 6900, (Mouse) 21367, (Rat) 25356

          View more View less

          Function(s)
          glycoprotein binding carbohydrate binding identical protein binding protein self-association protein binding
          Process(es)
          microtubule cytoskeleton organization cell adhesion cell-matrix adhesion axon guidance axonal fasciculation learning adult walking behavior positive regulation of protein processing cerebral cortex GABAergic interneuron migration central nervous system myelination regulation of axon diameter receptor internalization clustering of voltage-gated potassium channels negative regulation of neuron differentiation regulation of neuronal synaptic plasticity regulation of astrocyte differentiation positive regulation of adenosine receptor signaling pathway protein localization to juxtaparanode region of axon establishment of protein localization to juxtaparanode region of axon presynaptic membrane organization neuron migration axonogenesis regulation of cell morphogenesis involved in differentiation neuron projection development cellular protein localization
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5dd894dbdc-64gxr:80/100.66.134.75:80.
          git-commit: 83227db6267f00a4ced007f33c273793adf466ab
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0