PA1-30819 detects Carbonic Anhydrase II from rabbit, human samples.
PA1-30819 has been successfully used in Western blot applications.
The PA1-30819 immunogen is: Human Carbonic Anhydrase II, purified from human erythrocytes.
CA2 is one of several (at least 7) isozymes of carbonic anhydrase. Carbonic anhydrase catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis.