Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CTRL Antibodies

          Proteintech

          CTRL Polyclonal Antibody

          3 References
          View all (6) CTRL antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CTRL Polyclonal Antibody

          • Antibody Testing Data (6)
          CTRL Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CTRL Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CTRL Antibody (14856-1-AP) in ICC/IF

          Immunofluorescent analysis of BxPC-3 cells using 14856-1-AP (CTRL antibody) at dilution of 1:25 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CTRL Antibody in Immunocytochemistry (ICC/IF)
          CTRL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CTRL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CTRL Antibody in Western Blot (WB)
          CTRL Antibody in Western Blot (WB)
          CTRL Antibody in Western Blot (WB)

          Product Details

          14856-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:6,000
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          -

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CTRL Fusion Protein Ag6644 (1-264 aa encoded by BC063475)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.14 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MLLLSLTLS LVLLGSSWGC GIPAIKPALS FSQRIVNGEN AVLGSWPWQV SLQDSSGFHF CGGSLISQSW VVTAAHCNVS PGRHFVVLGE YDRSSNAEPL QVLSVSRAIT HPSWNSTTMN NDVTLLKLAS PAQYTTRISP VCLASSNEAL TEGLTCVTTG WGRLSGVGNV TPAHLQQVAL PLVTVNQCRQ YWGSSITDSM ICAGGAGASS CQGDSGGPLV CQKGNTWVLI GIVSWGTKNC NVRAPAVYTR VSKFSTWINQ VIAYN (1-264 aa encoded by BC063475)

          Target Information

          The CTRL gene encodes chymotrypsin-like protein, a serine protease involved in the digestion of proteins by cleaving peptide bonds. It belongs to the chymotrypsin family of enzymes that are predominantly expressed in the pancreas and play crucial roles in the breakdown of dietary proteins in the small intestine. CTRL functions by hydrolyzing specific peptide bonds, contributing to efficient protein digestion and nutrient absorption. This enzyme's activity is tightly regulated to prevent premature activation within the pancreas, which could lead to pancreatitis or other digestive disorders. The study of the CTRL gene is significant in understanding the mechanisms of digestive health and in diagnosing pancreatic disorders. Furthermore, CTRL's enzyme activity and its role in proteolytic pathways make it an important target for research in digestive enzyme supplementation therapies and treatment strategies for conditions involving impaired protein digestion.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: chymopasin; chymotrypsin A CTRA-1; chymotrypsin-like; Chymotrypsin-like protease CTRL-1

          View more View less

          Gene Aliases: 0910001G08Rik; 1810004D15Rik; AV005227; chymopasin; Ctra-1; Ctra1; CTRL; CTRL1; mFLJ00366

          View more View less

          UniProt ID: (Human) P40313

          View more View less

          Entrez Gene ID: (Human) 1506, (Rat) 117184, (Mouse) 109660

          View more View less

          Function(s)
          serine-type endopeptidase activity peptidase activity serine-type peptidase activity
          Process(es)
          proteolysis digestion
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0