Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LWRLQKLPA ELGPQLLHKI IDGICGRAYP VYQDYHTVWE SEEWMHVLED IAKFFKAIVG KNLPDEEIFQ QLNQLNSLHQ ETIMKCVKSR KDEIKQALSR EIVAISSAQL QDFDWQVKLA LSSDKIAALR MPLLSLHLDV KENGEVKPYS IEMSREELQN LIQSLEAANK VVLQL (9-182 aa encoded by BC008371)
The protein encoded by this gene binds coiled-coil domain-containing protein 22 (CCDC22), and this complex can regulate the turnover of I-kappa-B and the activation of NF-kappa-B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COMM domain-containing protein 8
Gene Aliases: COMMD8; MDS022
UniProt ID: (Human) Q9NX08
Entrez Gene ID: (Human) 54951
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support