PA5-29724 targets Collagen VI alpha 2 in WB applications and shows reactivity with Human samples.
The PA5-29724 immunogen is recombinant protein fragment corresponding to a region within amino acids 34 and 307 of Human COL6A2.
This gene encodes one of the three alpha chains of type VI collagen, a beaded filament collagen found in most connective tissues. The product of this gene contains several domains similar to von Willebrand Factor type A domains. These domains have been shown to bind extracellular matrix proteins, an interaction that explains the importance of this collagen in organizing matrix components. Mutations in this gene are associated with Bethlem myopathy and Ullrich scleroatonic muscular dystrophy. Three transcript variants have been identified for this gene.