Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • COG2 Antibodies

          Abnova

          COG2 Polyclonal Antibody, MaxPab™

          View all (9) COG2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite COG2 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          COG2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          COG2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          COG2 Antibody (H00022796-D01P) in WB

          Western Blot analysis of COG2 expression in transfected 293T cell line by COG2 MaxPab polyclonal antibody. Lane 1: COG2 transfected lysate(83.20 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          COG2 Antibody in Western Blot (WB)

          Product Details

          H00022796-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          COG2 (NP_031383.1, 1 a.a. approximately 738 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MEKSRMNLPK GPDTLCFDKD EFMKEDFDVD HFVSDCRKRV QLEELRDDLE LYYKLLKTAM VELINKDYAD FVNLSTNLVG MDKALNQLSV PLGQLREEVL SLRSSVSEGI RAVDERMSKQ EDIRKKKMCV LRLIQVIRSV EKIEKILNSQ SSKETSALEA SSPLLTGQIL ERIATEFNQL QFHAVQSKGM PLLDKVRPRI AGITAMLQQS LEGLLLEGLQ TSDVDIIRHC LRTYATIDKT RDAEALVGQV LVKPYIDEVI IEQFVESHPN GLQVMYNKLL EFVPHHCRLL REVTGGAISS EKGNTVPGYD FLVNSVWPQI VQGLEEKLPS LFNPGNPDAF HEKYTISMDF VRRLERQCGS QASVKRLRAH PAYHSFNKKW NLPVYFQIRF REIAGSLEAA LTDVLEDAPA ESPYCLLASH RTWSSLRRCW SDEMFLPLLV HRLWRLTLQI LARYSVFVNE LSLRPISNES PKEIKKPLVT GSKEPSITQG NTEDQGSGPS ETKPVVSISR TQLVYVVADL DKLQEQLPEL LEIIKPKLEM IGFKNFSSIS AALEDSQSSF SACVPSLSSK IIQDLSDSCF GFLKSALEVP RLYRRTNKEV PTTASSYVDS ALKPLFQLQS GHKDKLKQAI IQQWLEGTLS ESTHKYYETV SDVLNSVKKM EESLKRLKQA RKTTPANPVG PSGGMSDDDK IRLQLALDVE YLGEQIQKLG LQASDIKSFS ALAELVAAAK DQATAEQP

          Target Information

          The structure and function of the Golgi apparatus is controlled by a number of multi-protein complexes that are involved in glycosylation reactions and vesicular transport. The conserved oligomeric Golgi (COG) complex consists of three subcomplexes, termed LDLC, SEC34 and GTT (Golgi transport complex), all of which contain proteins necessary for proper Golgi operation. COG2 (conserved oligomeric Golgi complex subunit 2), also known as LDLC, is a 730 amino acid component of the COG complex. Localized to the cytoplasmic side of the Golgi apparatus, COG2 is required for proper Golgi morphology and function, specifically playing a role in Golgi ribbon formation and vesicular transport. Abnormal COG2 function may cause cell death, suggesting that COG2 is an important factor in cell viability.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: brefeldin A-sensitive, peripheral Golgi protein; COG 2; COG complex subunit 2; Component of oligomeric Golgi complex 2; conserved oligomeric Golgi complex protein 2; Conserved oligomeric Golgi complex subunit 2; low density lipoprotein receptor defect C complementing; Low density lipoprotein receptor defect C-complementing protein; Transcript ch4822

          View more View less

          Gene Aliases: COG2; LDLC

          View more View less

          UniProt ID: (Human) Q14746

          View more View less

          Entrez Gene ID: (Human) 22796

          View more View less

          Function(s)
          protein binding protein transporter activity protein complex binding
          Process(es)
          ER to Golgi vesicle-mediated transport intra-Golgi vesicle-mediated transport Golgi organization protein transport
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-7p7bh:80/100.66.135.138:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0