Recommended positive controls: Jurkat.
Predicted reactivity: Cow: 92%; Dog: 85%; Guinea Pig: 92%; Horse: 92%; Mouse: 92%; Rabbit: 92%; Rat: 92%.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CHX10 encodes a homeobox protein originally described as a retina-specific transcription factor. Mutations in this gene are associated with microphthalmia, cataracts and iris abnormalities.