FIGURE: 1 / 4
Product is shipped at room temperature as a lyophilized powder and should be stored at -20C upon receipt.
Reconstitute in 50 ul deionized water.
After binding acetylcholine, the AChR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.