FIGURE: 1 / 1
Immunogen sequence: MEGDGSDPEP PDAGEDSKSE NGENAPIYCI CRKPDINCFM IGCDNCNEWF HGDCIRITEK MAKAIREWYC RECREKDPKL EIRYRHKKSR ERDGNERDSS EPRDEGGGRK RPVPDPDLQR RAGSGTGVGA MLARGSASPH KSSPQPLVAT PSQHHQQQQQ QIKRSARMCG ECEACRRTED CGHCDFCRDM KKFGGPNKIR QKCRLRQCQL; Positive Samples: HepG2; Cellular Location: Nucleus speckle
Proteins that contain a CXXC motif within their DNA-binding domain, such as CXXC1, recognize CpG sequences and regulate gene expression (Carlone and Skalnik, 2001).