Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The antibody can recognize the isoform1 and isoform2.
CES7 is involved in the detoxification of xenobiotics and in the activation of ester and amide prodrugs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carboxylesterase 5; Carboxylesterase 5A; carboxylesterase 7; Carboxylesterase-like urinary excreted protein homolog; Cauxin; epididymis luminal protein 126
Gene Aliases: CAUXIN; CES4C1; CES5; CES5A; CES7; HEL126
UniProt ID: (Human) Q6NT32
Entrez Gene ID: (Human) 221223
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support