Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: KCPCDECVPL IKGAGIKQIY AGDVDVGKKK ADISYMRFGE LEGVSKFTWQ LNPSGAYGLE QNEPERRENG VLRPVPQKEE QHQDKKLRLG IH
May play an important role in testicular development and spermatogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytidine and dCMP deaminase domain-containing protein 1; Cytidine deaminase; protein kinase NYD-SP15; Testis development protein NYD-SP15
Gene Aliases: bA103J18.1; CDADC1; NYD-SP15
UniProt ID: (Human) Q9BWV3
Entrez Gene ID: (Human) 81602, (Rat) 361052
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support