Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AbboMax
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive control: Tonsil
Cellular location: Cell membrane.
CD72 is a transmembrane glycoprotein expressed as a homodimer especially in B cells, but also in other antigen presenting cells such as dendritic cells and macrophages. Through one of its immunoreceptor tyrosine-based inhibitory motives (ITIMs), CD72 interacts with tyrosine phosphatase SHP-1, thereby suppressing B cell responsiveness. Binding of CD72 with its ligand CD100 (Sema4D) prevents BCR association and phosphorylation of CD72 and results in dissociation of SHP-1 from CD72, thus enables B cell activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell differentiation antigen CD72; CD72; CD72 antigen; Lyb-2; RP11-331F9.9
Gene Aliases: CD72; CD72b; LYB2
UniProt ID: (Human) P21854
Entrez Gene ID: (Human) 971
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support