Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody is a monoclonal mixture of FITC-conjugated CD5 (clone M28623, isotype IgG2a) and PE-conjugated CD20 (clone DFA-7, isotype IgG1).
In humans, CD5 is a 67kDa T lymphocyte single chain transmembrane glycoprotein. It is present on all mature T lymphocytes, on most of thymocytes and on many T cell leukemias and lymphomas. It reacts with a subpopulation of activated B cells. The CD20 antigen is present on human pre-B lymphocytes and on B-lymphocytes atall stage of maturation, except on plasma cells. Low level expression of the CD20 antigen has been detected on normal T lymphocytes. CD20 is a non glycosylated protein with a molecular weight of 35 or 37 kDa depending on the degree of phosphorylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-lymphocyte antigen CD20; B-lymphocyte cell-surface antigen B1; B-lymphocyte surface antigen B1; Bp35; CD20; CD20 antigen; CD20 receptor; CD5; CD5 antigen (p56-62); CD5 antigen p56-62; Leukocyte surface antigen Leu-16; Lymphocyte antigen T1/Leu-1; Membrane-spanning 4-domains subfamily A member 1; membrane-spanning 4-domains, subfamily A, member 1; MGC3969; T-cell surface glycoprotein CD5
Gene Aliases: B1; Bp35; CD20; CD5; CVID5; LEU-16; LEU1; MS4A1; MS4A2; S7; T1
UniProt ID: (Human) P06127, (Human) P11836
Entrez Gene ID: (Human) 921, (Human) 931
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support