Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • TLR4 (CD284) Antibodies

          NeoBiotechnologies

          CD284 (Toll-Like Receptor 4) Monoclonal Antibody (TLR4/230)

          View all (75) TLR4 (CD284) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CD284 (Toll-Like Receptor 4) Monoclonal Antibody (TLR4/230)

          • Antibody Testing Data (1)
          CD284 (Toll-Like Receptor 4) Antibody in SDS-PAGE (SDS-PAGE)
          Group 53 Created with Sketch.
          CD284 (Toll-Like Receptor 4) Antibody in SDS-PAGE (SDS-PAGE)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD284 (Toll-Like Receptor 4) Antibody (7099-MSM1-P1ABX) in SDS-PAGE

          SDS-PAGE analysis Purified CD284 Mouse Monoclonal Antibody (TLR4/230). Confirmation of Integrity and Purity of Antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD284 (Toll-Like Receptor 4) Antibody in SDS-PAGE (SDS-PAGE)

          Product Details

          7099-MSM1-P1ABX

          Applications
          Tested Dilution
          Publications

          Functional Assay (Functional)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Dog, Guinea pig, Human, Non-human primate, Pig, Rat

          Host/Isotype

          Mouse / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          TLR4/230

          Immunogen

          Recombinant full-length human TLR4 protein
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A/G

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          TLR4 is member of the Toll like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. This receptor is most abundantly expressed in placenta, and in myelomonocytic subpopulation of the leukocytes. Mammalian cells respond to LPS by activating TLR4. TLR4 belongs to the multi-protein complex of lipopolysaccharide (LPS) receptor, containing CD14, LY96 and TLR4, and is involved in signal transduction events induced by lipopolysaccharide (LPS) found in most gram-negative bacteria. TLR4 aids in the recognition of pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. Mutations in the TLR4 gene have been associated with differences in LPS responsiveness. Also, several transcript variants of the TLR4 gene have been found, but the protein coding potential of most of them is uncertain. TLR4 is expressed by peripheral blood monocytes and a small population of B-cells and is also expressed in human placenta. Studies with TLR4-deficient mice indicate that the main ligand for TLR is lipopolysaccharide. Consequently, these mice also showed increased susceptibility to Gram-negative sepsis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD284; homolog of Drosophila toll; hToll; Toll-like receptor 4; Toll-like receptor4 protein; Toll4

          View more View less

          Gene Aliases: ARMD10; CD284; TLR-4; TLR4; TOLL

          View more View less

          UniProt ID: (Human) O00206, (Rat) Q9QX05

          View more View less

          Entrez Gene ID: (Pig) 399541, (Human) 7099, (Dog) 403417, (Rat) 29260

          View more View less

          Function(s)
          lipopolysaccharide binding lipopolysaccharide receptor activity receptor activity transmembrane signaling receptor activity protein binding phosphatidylinositol 3-kinase binding
          Process(es)
          activation of MAPK activity toll-like receptor signaling pathway B cell proliferation involved in immune response nitric oxide production involved in inflammatory response regulation of dendritic cell cytokine production MyD88-dependent toll-like receptor signaling pathway MyD88-independent toll-like receptor signaling pathway inflammatory response immune response I-kappaB kinase/NF-kappaB signaling I-kappaB phosphorylation positive regulation of platelet activation positive regulation of gene expression astrocyte development detection of fungus positive regulation of B cell proliferation lipopolysaccharide-mediated signaling pathway response to lipopolysaccharide detection of lipopolysaccharide interferon-gamma production negative regulation of interferon-gamma production negative regulation of interleukin-17 production negative regulation of interleukin-23 production negative regulation of interleukin-6 production negative regulation of tumor necrosis factor production positive regulation of chemokine production positive regulation of interferon-alpha production positive regulation of interferon-beta production positive regulation of interferon-gamma production positive regulation of interleukin-1 production positive regulation of interleukin-10 production positive regulation of interleukin-12 production positive regulation of interleukin-6 production positive regulation of interleukin-8 production positive regulation of tumor necrosis factor production negative regulation of MyD88-independent toll-like receptor signaling pathway toll-like receptor 4 signaling pathway TRIF-dependent toll-like receptor signaling pathway T-helper 1 type immune response macrophage activation positive regulation of NF-kappaB import into nucleus positive regulation of tumor necrosis factor biosynthetic process defense response to bacterium positive regulation of interleukin-12 biosynthetic process innate immune response positive regulation of MHC class II biosynthetic process positive regulation of interferon-beta biosynthetic process positive regulation of interleukin-8 biosynthetic process positive regulation of nitric oxide biosynthetic process negative regulation of osteoclast differentiation positive regulation of transcription from RNA polymerase II promoter positive regulation of JNK cascade interleukin-1 beta secretion regulation of cytokine secretion positive regulation of inflammatory response defense response to Gram-negative bacterium positive regulation of NF-kappaB transcription factor activity positive regulation of nitric-oxide synthase biosynthetic process intestinal epithelial structure maintenance positive regulation of macrophage cytokine production necroptotic process negative regulation of ERK1 and ERK2 cascade positive regulation of ERK1 and ERK2 cascade positive regulation of nucleotide-binding oligomerization domain containing 1 signaling pathway positive regulation of nucleotide-binding oligomerization domain containing 2 signaling pathway cellular response to lipopolysaccharide cellular response to lipoteichoic acid cellular response to mechanical stimulus apoptotic signaling pathway positive regulation of NLRP3 inflammasome complex assembly response to hypoxia microglial cell activation activation of innate immune response innate immune response-activating signal transduction skeletal muscle contraction leukotriene metabolic process response to oxidative stress response to bacterium response to toxic substance response to activity immunoglobulin mediated immune response response to retinoic acid response to progesterone response to insulin positive regulation of stress-activated MAPK cascade positive regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling response to morphine positive regulation of DNA binding response to ethanol positive regulation of lymphocyte proliferation regulation of inflammatory response positive regulation of peptidyl-tyrosine phosphorylation positive regulation of sequence-specific DNA binding transcription factor activity response to glucocorticoid regulation of sensory perception of pain response to fatty acid cellular response to retinoic acid response to gold nanoparticle
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66966984d8-2vwff:80/100.66.134.75:80.
          git-commit: 4f7892df6eac4c3aaa23131f1084e28be0ddeb54
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0