Product References
Advanced immunophenotyping: A powerful tool for immune profiling, drug screening, and a personalized treatment approach.
Frontiers in immunology
Preglej T,Brinkmann M,Steiner G,Aletaha D,Göschl L,Bonelli M
17-0259-42 was used in Flow cytometry/Cell sorting to investigate the potential of multi-parameter spectral flow cytometry for the identification of patients suffering from autoimmune diseases and its power as an evaluation tool for in vitro drug screening approaches (advanced immunophenotyping).
Tue Apr 11 00:00:00 UTC 2023
BLIMP1 and NR4A3 transcription factors reciprocally regulate antitumor CAR T cell stemness and exhaustion.
Science translational medicine
Jung IY,Narayan V,McDonald S,Rech AJ,Bartoszek R,Hong G,Davis MM,Xu J,Boesteanu AC,Barber-Rotenberg JS,Plesa G,Lacey SF,Jadlowsky JK,Siegel DL,Hammill DM,Cho-Park PF,Berger SL,Haas NB,Fraietta JA
17-0259-42 was used in Flow cytometry/Cell sorting to investigate the dual targeting of PRDM1 and NR4A3 as a promising approach to advance adoptive cell immuno-oncotherapy.
Wed Nov 09 00:00:00 UTC 2022
EC-18 prevents autoimmune arthritis by suppressing inflammatory cytokines and osteoclastogenesis.
Arthritis research & therapy
Park JS,Yang SC,Jeong HY,Lee SY,Ryu JG,Choi JW,Kang HY,Kim SM,Hwang SH,Cho ML,Park SH
17-0259-42 was used in Flow cytometry/Cell sorting to investigate the efficacy of EC-18 on the development of RA.
Thu Nov 17 00:00:00 UTC 2022
Ageing promotes early T follicular helper cell differentiation by modulating expression of RBPJ.
Aging cell
Webb LMC,Fra-Bido S,Innocentin S,Matheson LS,Attaf N,Bignon A,Novarino J,Fazilleau N,Linterman MA
17-0259-42 was used in Flow Cytometry to show increased formation of Tfh precursors (pre-Tfh) but no associated increase in germinal centre (GC)-Tfh cells in aged mice, suggesting age-driven promotion of only early Tfh cell differentiation.
Fri Jan 01 00:00:00 UTC 2021
Taurine promotes the production of CD4+CD25+FOXP3+ Treg cells through regulating IL-35/STAT1 pathway in a mouse allergic rhinitis model.
Allergy, asthma, and clinical immunology : official journal of the Canadian Society of Allergy and Clinical Immunology
Zhou J,Lu Y,Wu W,Feng Y
17-0259-42 was used in Flow Cytometry to show that the restoration of Treg populations by taurine normalizes the inflammatory response, reduces AR symptomology, and reduces histopathologic signs of AR.
Sat Jun 19 00:00:00 UTC 2021
Biological and molecular studies on specific immune cells treated with checkpoint inhibitors for the thera-personal approach of breast cancer patients (ex-vivo study).
Oncology research
El-Houseini ME,Arafat MS,El-Husseiny AM,Kasem IM,Kamel MM,El-Habashy AH,Khafagy MM,Radwan EM,Helal MH,Abdellateif MS
17-0259-42 was used in Flow Cytometry to optimize dendritic cell (DC)-based immunotherapy for breast cancer (BC) by co-culturing immune cells (DCs, T cells, tumor-infiltrating lymphocytes, and tumor-infiltrating DCs) treated with anti-PD1 and anti-CTLA4 monoclonal antibodies with autologous breast cancer cells (BCCs), leading to significant upregulation of activation markers on immune cells, downregulation of regulatory T cell markers and key cancer-related genes in BCCs, and changes in protein
Tue Jun 13 00:00:00 UTC 2023
Superoxide Dismutase 3 Controls the Activation and Differentiation of CD4+T Cells.
Frontiers in immunology
Agrahari G,Sah SK,Bang CH,Kim YH,Kim TY
17-0259-42 was used in Flow Cytometry to show that SOD3 can attenuate CD4+T cell activation through modulation of the downstream signalings and restrict CD4+T cell differentiation.
Tue Aug 03 00:00:00 UTC 2021
Metabolic Disturbance and Th17/Treg Imbalance Are Associated With Progression of Gingivitis.
Frontiers in immunology
Wang W,Wang X,Lu S,Lv H,Zhao T,Xie G,Du Y,Fan Y,Xu L
17-0259-42 was used in Flow cytometry/Cell sorting to explore the role of metabolic disturbance in immunoregulation of gingivitis targeting T helper 17 cells/regulatory T cells.
Wed Oct 20 00:00:00 UTC 2021
Propionic Acid Shapes the Multiple Sclerosis Disease Course by an Immunomodulatory Mechanism.
Cell
Duscha A,Gisevius B,Hirschberg S,Yissachar N,Stangl GI,Dawin E,Bader V,Haase S,Kaisler J,David C,Schneider R,Troisi R,Zent D,Hegelmaier T,Dokalis N,Gerstein S,Del Mare-Roumani S,Amidror S,Staszewski O,Poschmann G,Stühler K,Hirche F,Balogh A,Kempa S,Träger P,Zaiss MM,Holm JB,Massa MG,Nielsen HB,Faissner A,Lukas C,Gatermann SG,Scholz M,Przuntek H,Prinz M,Forslund SK,Winklhofer KF,Müller DN,Linker RA,Gold R,Haghikia A
17-0259 was used in Flow cytometry/Cell sorting to investigate propionic acid (PA) in multiple sclerosis (MS), an autoimmune and neurodegenerative disease.
Thu Mar 19 00:00:00 UTC 2020
FOXO1 promotes HIV latency by suppressing ER stress in T cells.
Nature microbiology
Vallejo-Gracia A,Chen IP,Perrone R,Besnard E,Boehm D,Battivelli E,Tezil T,Krey K,Raymond KA,Hull PA,Walter M,Habrylo I,Cruz A,Deeks S,Pillai S,Verdin E,Ott M
17-0259 was used in Flow cytometry/Cell sorting to uncover a link of FOXO1, ER stress and HIV infection that could be therapeutically exploited to selectively reverse T-cell quiescence and reduce the size of the latent viral reservoir.
Tue Sep 01 00:00:00 UTC 2020
T-Lymphocyte Subset Distribution and Activity in Patients With Glaucoma.
Investigative ophthalmology & visual science
Yang X,Zeng Q,Göktas E,Gopal K,Al-Aswad L,Blumberg DM,Cioffi GA,Liebmann JM,Tezel G
17-0259-42 was used in Flow Cytometry to analyse T lymphocytes in patients’ blood, to explore the T-cell– mediated component of systemic immunity.
Fri Mar 01 00:00:00 UTC 2019
Cell surface cathepsin G can be used as an additional marker to distinguish T cell subsets.
Biomedical reports
Penczek A,Burster T
17-0259 was used in Flow cytometry/Cell sorting to analyse peripheral blood mononuclear cells to characterise T cell populations in terms of their cathepsin G content by flow cytometry.
Mon Apr 01 00:00:00 UTC 2019
A natural killer-dendritic cell axis defines checkpoint therapy-responsive tumor microenvironments.
Nature medicine
Barry KC,Hsu J,Broz ML,Cueto FJ,Binnewies M,Combes AJ,Nelson AE,Loo K,Kumar R,Rosenblum MD,Alvarado MD,Wolf DM,Bogunovic D,Bhardwaj N,Daud AI,Ha PK,Ryan WR,Pollack JL,Samad B,Asthana S,Chan V,Krummel MF
17-0259 was used in Flow cytometry/Cell sorting to reveal that innate immune stimulatory dendritic cells and natural killer cells cluster together as an excellent prognostic tool for T cell-directed immunotherapy and that these innate cells are necessary for enhanced T cell tumor responses, suggesting this axis as a target for new therapies.
Wed Aug 01 00:00:00 UTC 2018
Metabolic reprogramming of human CD8+ memory T cells through loss of SIRT1.
The Journal of experimental medicine
Jeng MY,Hull PA,Fei M,Kwon HS,Tsou CL,Kasler H,Ng CP,Gordon DE,Johnson J,Krogan N,Verdin E,Ott M
17-0259-42 was used in Flow Cytometry to identify the evolutionarily conserved SIRT1-FoxO1 axis as a regulator of resting CD8+ memory T cell metabolism and activity in humans.
Tue Jan 02 00:00:00 UTC 2018
The Genetic Polymorphisms of NLRP3 Inflammasome Associated with T Helper Cells in Patients with Multiple Myeloma.
Journal of immunology research
Zhao X,Hua M,Yan S,Yu J,Han F,Zhong C,Wang R,Zhang C,Hou M,Ma D
Fri Nov 09 00:00:00 UTC 2018
Small Molecule Targeting of Specific BAF (mSWI/SNF) Complexes for HIV Latency Reversal.
Cell chemical biology
Marian CA,Stoszko M,Wang L,Leighty MW,de Crignis E,Maschinot CA,Gatchalian J,Carter BC,Chowdhury B,Hargreaves DC,Duvall JR,Crabtree GR,Mahmoudi T,Dykhuizen EC
17-0259-42 was used in Flow cytometry/Cell sorting to report a high-throughput screen for inhibitors of BAF-mediated transcription in cells and the subsequent identification of a 12-membered macrolactam.
Thu Dec 20 00:00:00 UTC 2018
Mesenchymal stromal cells from myelodysplastic and acute myeloid leukemia patients display in vitro reduced proliferative potential and similar capacity to support leukemia cell survival.
Stem cell research & therapy
Corradi G,Baldazzi C,Očadlíková D,Marconi G,Parisi S,Testoni N,Finelli C,Cavo M,Curti A,Ciciarello M
17-0259 was used in Flow cytometry/Cell sorting to compare mesenchymal stromal cells isolated from healthy, myelodysplastic syndrome, and acute myeloid leukaemia subjects to investigate MSC alterations and to emphasise putative common and/or diverse features.
Thu Oct 25 00:00:00 UTC 2018
Targeting a CAR to the TRAC locus with CRISPR/Cas9 enhances tumour rejection.
Nature
Eyquem J,Mansilla-Soto J,Giavridis T,van der Stegen SJ,Hamieh M,Cunanan KM,Odak A,Gönen M,Sadelain M
17-0259 was used in Flow cytometry/Cell sorting to demonstrate that targeting a chimeric antigen receptor coding sequence to the TCR locus increases the therapeutic potency of engineered T cells.
Thu Mar 02 00:00:00 UTC 2017
SMYD2-Mediated Histone Methylation Contributes to HIV-1 Latency.
Cell host & microbe
Boehm D,Jeng M,Camus G,Gramatica A,Schwarzer R,Johnson JR,Hull PA,Montano M,Sakane N,Pagans S,Godin R,Deeks SG,Krogan NJ,Greene WC,Ott M
17-0259 was used in Flow cytometry/Cell sorting to propose that a SMYD2-H4K20me1-L3MBTL1 axis contributes to HIV-1 latency and can be targeted with small-molecule SMYD2 inhibitors.
Wed May 10 00:00:00 UTC 2017
Erythropoietin Receptor-Mediated Molecular Crosstalk Promotes T Cell Immunoregulation and Transplant Survival.
Journal of the American Society of Nephrology : JASN
Purroy C,Fairchild RL,Tanaka T,Baldwin WM,Manrique J,Madsen JC,Colvin RB,Alessandrini A,Blazar BR,Fribourg M,Donadei C,Maggiore U,Heeger PS,Cravedi P
17-0259 was used in Flow cytometry/Cell sorting to delineate the protolerogenic properties of EPO in inhibiting conventional T cells while simultaneously promoting Treg induction.
Tue Aug 01 00:00:00 UTC 2017
HIV-1 competition experiments in humanized mice show that APOBEC3H imposes selective pressure and promotes virus adaptation.
PLoS pathogens
Nakano Y,Misawa N,Juarez-Fernandez G,Moriwaki M,Nakaoka S,Funo T,Yamada E,Soper A,Yoshikawa R,Ebrahimi D,Tachiki Y,Iwami S,Harris RS,Koyanagi Y,Sato K
17-0259 was used in Flow cytometry/Cell sorting to determine the anti-viral effect of endogenous A3H in vivo.
Mon May 01 00:00:00 UTC 2017
Antigen-Presenting Human γδ T Cells Promote Intestinal CD4+ T Cell Expression of IL-22 and Mucosal Release of Calprotectin.
Journal of immunology (Baltimore, Md. : 1950)
Tyler CJ,McCarthy NE,Lindsay JO,Stagg AJ,Moser B,Eberl M
17-0259 was used in Flow cytometry/Cell sorting to study the role of γδ T cells in the regulation of IL-22 expression.
Mon May 01 00:00:00 UTC 2017
Differences in Peripheral Blood Lymphocytes between Brand-Name and Generic Tacrolimus Used in Stable Liver Transplant Recipients.
Medical principles and practice : international journal of the Kuwait University, Health Science Centre
Kim JM,Kwon CHD,Joh JW,Sinn DH,Choi GS,Park JB,Kang ES,Lee SK
17-0259 was used in Flow cytometry/Cell sorting to show that brand-name tacrolimus could have more potential immunosuppressive activity than generic tacrolimus regarding the contribution of CD4+Foxp3+ T cells to graft tolerance in liver transplant recipients.
Mon Mar 19 00:00:00 UTC 2018
Oroxylin A inhibits the generation of Tregs in non-small cell lung cancer.
Oncotarget
Shen L,Zhang LL,Li H,Liu X,Yu XX,Hu P,Hui H,Guo QL,Zhang S
17-0259 was used in Flow cytometry/Cell sorting to demonstrate that oroxylin A inhibits Treg generation in lung cancer by inhibiting TGF-β1 response and secretion via NF-κB signalling.
Tue Jul 25 00:00:00 UTC 2017
Reciprocal regulation of the Il9 locus by counteracting activities of transcription factors IRF1 and IRF4.
Nature communications
Campos Carrascosa L,Klein M,Kitagawa Y,Lückel C,Marini F,König A,Guralnik A,Raifer H,Hagner-Benes S,Rädler D,Böck A,Kang C,Lohoff M,Garn H,Schaub B,Berberich-Siebelt F,Sakaguchi S,Bopp T,Huber M
17-0259 was used in Flow cytometry/Cell sorting to study the opposing effects of interferon regulatory factor1 and IRF4 on T helper 9 cell differentiation.
Fri May 12 00:00:00 UTC 2017
Function of Treg Cells Decreased in Patients With Systemic Lupus Erythematosus Due To the Effect of Prolactin.
Medicine
Legorreta-Haquet MV,Chávez-Rueda K,Chávez-Sánchez L,Cervera-Castillo H,Zenteno-Galindo E,Barile-Fabris L,Burgos-Vargas R,Álvarez-Hernández E,Blanco-Favela F
17-0259 was used in Flow cytometry/Cell sorting to investigate the role of prolactin in Treg-mediated immune response, showing that it decreased Treg function in patients with systemic Lupus Erythematosus.
Mon Feb 01 00:00:00 UTC 2016
Reply to Suppressive activity of human regulatory T cells is maintained in the presence of TNF.
Nature medicine
Nie H,Zheng Y,Li R,Zhang J
17-0259-42 was used in flow cytometry to show that reply to suppressive activity of human regulatory T cells is maintained in the presence of TNF.
Fri Jan 01 00:00:00 UTC 2016
Durable complete responses off all treatment in patients with metastatic malignant melanoma after sequential immunotherapy followed by a finite course of BRAF inhibitor therapy.
Cancer biology & therapy
Wyluda EJ,Cheng J,Schell TD,Haley JS,Mallon C,Neves RI,Robertson G,Sivik J,Mackley H,Talamo G,Drabick JJ
17-0259 was used in Flow cytometry/Cell sorting to study the effectiveness of different therapy regimes on treating melanoma after failure of immunotherapy.
Tue Mar 01 00:00:00 UTC 2016
Vitamin D deficiency contributes to the reduction and impaired function of naïve CD45RA⁺ regulatory T cell in chronic heart failure.
Journal of immunology research
Ma YH,Zhou YL,Yue CY,Zhang GH,Deng L,Xie GH,Xu WP,Shen LS
17-0259 was used in Flow cytometry/Cell sorting to investigate the impact of vitamin D supplementation on naïve Tregs.
Tue Jan 26 00:00:00 UTC 2016
Statins increase the frequency of circulating CD4+ FOXP3+ regulatory T cells in healthy individuals.
Journal of immunology research
Rodríguez-Perea AL,Montoya CJ,Olek S,Chougnet CA,Velilla PA
17-0259 was used in Flow cytometry/Cell sorting to investigate the effect of statins on Tregs in the absence of inflammation, showing that they increase the frequency of CD4(+)FOXP3(+) Tregs.
Tue Dec 01 00:00:00 UTC 2015
Skewed T-helper (Th)1/2- and Th17/T regulatory‑cell balances in patients with renal cell carcinoma.
Molecular medicine reports
Li L,Yang C,Zhao Z,Xu B,Zheng M,Zhang C,Min Z,Guo J,Rong R
17-0259 was used in Flow cytometry/Cell sorting to demonstrate that the balance of Th1 and Th2 cells is skewed towards the Th2 profile in the peripheral blood of patients with RCC.
Sun Feb 01 00:00:00 UTC 2015
A Phenotypic Analysis of Regulatory T Cells and Uterine NK Cells from First Trimester Pregnancies and Associations with HLA-G.
American journal of reproductive immunology (New York, N.Y. : 1989)
Djurisic S,Skibsted L,Hviid TV
17-0259-42 was used in Flow cytometry/Cell sorting to conclude that the NK subset in the placental bed displays a unique phenotype that may be influenced by mHLA-G on trophoblast cells and locally accumulated sHLA-G in the uterus.
Sun Nov 01 00:00:00 UTC 2015
Depressive symptoms post hip fracture in older adults are associated with phenotypic and functional alterations in T cells.
Immunity & ageing : I & A
Duggal NA,Upton J,Phillips AC,Hampson P,Lord JM
17-0259 was used in Flow cytometry/Cell sorting to investigate the synergistic effects of hip fracture physical trauma and psychological depressive symptoms on the aged immune system.
Wed Jan 28 00:00:00 UTC 2015
A multiple sclerosis-associated variant of CBLB links genetic risk with type I IFN function.
Journal of immunology (Baltimore, Md. : 1950)
Stürner KH,Borgmeyer U,Schulze C,Pless O,Martin R
17-0259 was used in Flow cytometry/Cell sorting to suggest a role of the CBLB rs12487066 variant in the interactions of a genetic risk factor and IFN function during viral infections in MS.
Sat Nov 01 00:00:00 UTC 2014
CD1d(hi)CD5+ B cells expanded by GM-CSF in vivo suppress experimental autoimmune myasthenia gravis.
Journal of immunology (Baltimore, Md. : 1950)
Sheng JR,Quan S,Soliven B
17-0259 was used in Flow cytometry/Cell sorting to investigate the mechanisms underlying the tolorogenic effects of B10 cells in Experimental Autoimmune Myasthenia Gravis.
Mon Sep 15 00:00:00 UTC 2014
Multiple sclerosis-associated IL2RA polymorphism controls GM-CSF production in human TH cells.
Nature communications
Hartmann FJ,Khademi M,Aram J,Ammann S,Kockum I,Constantinescu C,Gran B,Piehl F,Olsson T,Codarri L,Becher B
17-0259 was used in Flow cytometry/Cell sorting to study the link between granulocyte-macrophage colony-stimulating factor release TH cells and interleukin 2 signalling in the context of multiple sclerosi.
Fri Oct 03 00:00:00 UTC 2014
Quantitative reconstruction of leukocyte subsets using DNA methylation.
Genome biology
Accomando WP,Wiencke JK,Houseman EA,Nelson HH,Kelsey KT
17-0259 was used in Flow cytometry/Cell sorting to study how precise immune cell differential estimates can be reconstructed using only DNA rather than whole cells.
Wed Mar 05 00:00:00 UTC 2014
Fully human antagonistic antibodies against CCR4 potently inhibit cell signaling and chemotaxis.
PloS one
Hagemann UB,Gunnarsson L,Géraudie S,Scheffler U,Griep RA,Reiersen H,Duncan AR,Kiprijanov SM
17-0259 was used in Flow cytometry/Cell sorting to demonstrate a significant survival benefit for animals treated with newly selected anti-CCR4 antibodies.
Fri Nov 13 00:00:00 UTC 2015
Attenuated antigen-specific T cell responses in cirrhosis are accompanied by elevated serum interleukin-10 levels and down-regulation of HLA-DR on monocytes.
BMC gastroenterology
Peter J,Frey O,Stallmach A,Bruns T
17-0259 was used in Flow cytometry/Cell sorting to investigate defects in cellular immunity in cirrhosis.
Wed Feb 27 00:00:00 UTC 2013
Dual role of miR-21 in CD4+ T-cells: activation-induced miR-21 supports survival of memory T-cells and regulates CCR7 expression in naive T-cells.
PloS one
Smigielska-Czepiel K,van den Berg A,Jellema P,Slezak-Prochazka I,Maat H,van den Bos H,van der Lei RJ,Kluiver J,Brouwer E,Boots AM,Kroesen BJ
17-0259 was used in Flow cytometry/Cell sorting to investigate the contribution of miR-21 to immune cell function.
Mon Jul 28 00:00:00 UTC 2014
Elevated levels of CD4(+)CD25(+)FoxP3(+) T cells in systemic sclerosis patients contribute to the secretion of IL-17 and immunosuppression dysfunction.
PloS one
Liu X,Gao N,Li M,Xu D,Hou Y,Wang Q,Zhang G,Sun Q,Zhang H,Zeng X
17-0259 was used in Flow cytometry/Cell sorting to investigate the role of Treg subsets in the immune imbalance in naive systemic sclerosis (SSc), showing that elevated levels of CD4(+)CD25(+)FoxP3(+) T cells contribute to the secretion of IL-17 and immunosuppression dysfunction.
Thu Jan 16 00:00:00 UTC 2014
Induction of endometriosis alters the peripheral and endometrial regulatory T cell population in the non-human primate.
Human reproduction (Oxford, England)
Braundmeier A,Jackson K,Hastings J,Koehler J,Nowak R,Fazleabas A
17-0259 was used in Flow cytometry/Cell sorting to study how inducing endometriosis can alter the regulatory T cell population in the non-human primate.
Fri Jun 01 00:00:00 UTC 2012
Antigen-specific IL-2 secretion correlates with NK cell responses after immunization of Tanzanian children with the RTS,S/AS01 malaria vaccine.
Journal of immunology (Baltimore, Md. : 1950)
Horowitz A,Hafalla JC,King E,Lusingu J,Dekker D,Leach A,Moris P,Cohen J,Vekemans J,Villafana T,Corran PH,Bejon P,Drakeley CJ,von Seidlein L,Riley EM
17-0259 was used in Flow cytometry/Cell sorting to analyse immune cell responses to RTS,S and P. falciparum circumsporozoite protein component antigens in Tanzanian children.
Tue May 15 00:00:00 UTC 2012
Prolactin down-regulates CD4+CD25hiCD127low/- regulatory T cell function in humans.
Journal of molecular endocrinology
Legorreta-Haquet MV,Chávez-Rueda K,Montoya-Díaz E,Arriaga-Pizano L,Silva-García R,Chávez-Sánchez L,Moreno-Lafont M,Zenteno-Galindo E,Blanco-Favela F
17-0259 was used in Flow cytometry/Cell sorting to investigate the mechanisms by which prolactin regulates Tregs, showing that it down-regulates CD4+CD25hiCD127low/- regulatory T cell function.
Wed Feb 01 00:00:00 UTC 2012
Epigenetic modification of the human CCR6 gene is associated with stable CCR6 expression in T cells.
Blood
Steinfelder S,Floess S,Engelbert D,Haeringer B,Baron U,Rivino L,Steckel B,Gruetzkau A,Olek S,Geginat J,Huehn J,Hamann A
17-0259 was used in Flow cytometry/Cell sorting to identify a noncoding region of the human CCR6 gene with methylation-sensitive transcriptional activity in CCR6(+) T cells that controls stable CCR6 expression via epigenetic mechanisms.
Thu Mar 10 00:00:00 UTC 2011
Anticardiac myosin immunity and chronic allograft vasculopathy in heart transplant recipients.
Journal of immunology (Baltimore, Md. : 1950)
Kalache S,Dinavahi R,Pinney S,Mehrotra A,Cunningham MW,Heeger PS
17-0259 was used in Flow cytometry/Cell sorting to investigate the pathogenesis of chronic allograft vasculopathy in heart transplant failure.
Fri Jul 15 00:00:00 UTC 2011
Fms-like tyrosine kinase 3 ligand decreases T helper type 17 cells and suppressors of cytokine signaling proteins in the lung of house dust mite-sensitized and -challenged mice.
American journal of respiratory cell and molecular biology
McGee HS,Stallworth AL,Agrawal T,Shao Z,Lorence L,Agrawal DK
17-0259 was used in Flow cytometry/Cell sorting to evaluate the effect of Flt3-L on Th17 cells and expression of suppressors of cytokine signaling proteins, showing that both are decreased.
Mon Nov 01 00:00:00 UTC 2010
The rapid induction of HLA-E is essential for the survival of antigen-activated naive CD4 T cells from attack by NK cells.
Journal of immunology (Baltimore, Md. : 1950)
Takao S,Ishikawa T,Yamashita K,Uchiyama T
17-0259 was used in Flow cytometry/Cell sorting to investigate the mechanisms by which NK cells regulate adaptive immunity, showing that rapid induction of HLA-E is essential for the survival of antigen-activated naive CD4 T cells.
Mon Nov 15 00:00:00 UTC 2010
Expression and function of PDCD1 at the human maternal-fetal interface.
Biology of reproduction
Taglauer ES,Trikhacheva AS,Slusser JG,Petroff MG
17-0259 was used in Flow cytometry/Cell sorting to examine whether human maternal T cells express PDCD1.
Mon Sep 01 00:00:00 UTC 2008