Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Keep as concentrated solution.
Predicted reactivity: Mouse (83%), Rat (84%), Rhesus Monkey (99%), Chimpanzee (99%), Bovine (91%).
Positive Control: HepG2.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CCDC92 (Coiled-Coil Domain Containing 92) is a Protein Coding gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil domain-containing protein 92; limkain beta 2; Limkain beta-2
Gene Aliases: CCDC92
UniProt ID: (Human) Q53HC0
Entrez Gene ID: (Human) 80212
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support