Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MGCRHSRLSS CKPPKKKRQE PEPEQPPRPE PHELGPLNGD TAITVQLCAS EEAERHQKDI TRILQQHEEE KKKWAQQVEK ERELELRDRL DEQQRVLEGK NEEALQVLRA SYEQEKEALT HSFREASSTQ QETIDRLTSQ LEAFQAKMKR VEESILSRNY KKHIQDYGSP SQFWEQELES LHFVIEMKNE RIHELDRRLI LMETVKEKNL ILEEKITTLQ QENEDLHVRS RNQVVLSRQL SEDLLLTREA LEKEVQLRRQ LQQEKEELLY RVLGANASPA FPLAPVTPTE VSFLAT
May act as a scaffold to regulate the recruitment and assembly of spindle midzone components. Required for the localization of AURKB and PLK1 to the spindle midzone. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil domain-containing protein 69
Gene Aliases: CCDC69
UniProt ID: (Human) A6NI79
Entrez Gene ID: (Human) 26112
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support