Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MCAASASPDN LVFHMKNEMR NIKYKPVDYQ QLRALTEAKK LASASAKLKI RKAMLTSKLS KEQTLIKQHK QVWWQEYQRL NEVRCKMESE IKSLLNEENI GNECLCDLTN FEQELSEQQC TYLKNVINPI QQLRADLKYR QHHTLQHSHP HIEFNSVKVL EEVDFVKKQL KTVFERLRLE QQRIENDLSD WSIKILDHSL EEKTNPLSEL PIELESLECP YPDLKSSILS EFYKFTQKYQ KKLQDFNLQL EDIYR
CCDC148 (Coiled-Coil Domain Containing 148) is a Protein Coding gene. Diseases associated with CCDC148 include Vulva Cancer and Vulvar Disease. An important paralog of this gene is CCDC112. [GeneCards]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil domain-containing protein 148
Gene Aliases: CCDC148
UniProt ID: (Human) Q8NFR7
Entrez Gene ID: (Human) 130940
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support