Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAALDLRAE LDSLVLQLLG DLEELEGKRT VLNARVEEGW LSLAKARYAM GAKSVGPLQY ASHMEPQVCL HASEAQEGLQ KFKVVRAGVH APEEVGPREA GLRRRKGPTK TPEPESSEAP QDPLNWFGIL VPHSLRQAQA SFRDGLQLAA DIASLQNRID WGRSQLRGLQ EKLKQLEPGA A (1-180 aa encoded by BC006429)
The CCDC115 gene encodes a protein known as coiled-coil domain-containing protein 115, which is involved in cellular processes related to membrane trafficking and Golgi apparatus function. CCDC115 is integral to maintaining Golgi homeostasis and is implicated in the functioning of the vacuolar H+-ATPase (V-ATPase) proton pump, critical for acidifying intracellular compartments. Mutations in CCDC115 have been associated with a genetic condition known as congenital disorder of glycosylation type IIo (CDG-IIo), which leads to defects in protein glycosylation. This disorder results in a spectrum of symptoms, including developmental delays, neurological impairments, and multi-systemic dysfunction due to improper glycosylation of proteins, impacting their stability and function. Research indicates that CCDC115's activity in ion transport and vacuolar acidification is essential for Golgi apparatus function and overall cellular homeostasis. Investigations into CCDC115 are crucial for understanding its role in glycosylation-related disorders and for developing potential therapeutic strategies to manage these conditions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ccp1; Coiled-coil domain-containing protein 115; Coiled-coil protein 1; Vacuolar ATPase assembly protein VMA22
Gene Aliases: 2310061I09Rik; CCDC115; Ccp1; CDG2O; RGD1304653; VMA22
UniProt ID: (Human) Q96NT0, (Mouse) Q8VE99
Entrez Gene ID: (Human) 84317, (Rat) 363213, (Mouse) 69668
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support