FIGURE: 1 / 6
PA5-31565 targets C9orf72 in IHC (P), ICC/IF, and WB applications and shows reactivity with Human and mouse samples.
The PA5-31565 immunogen is recombinant fragment corresponding to a region within amino acids 1 and 198 of Human C9orf72.
C9orf72 may play a role in endosomal trafficking and autophagy. It is known to interact with HNRNPA1, HNRNPA2B1, UBQLN2, and small Rab GTPSases. C9orf72 localizes in the nucleus, cytoplasm, endosome, lysosome, and autophagosome. Mutations in this gene can result in frontotemporal dementia and/or amyotrophic lateral sclerosis.