Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAGMVDFQD EEQVKSFLEN MEVECNYHCY HEKDPDGCYR LVDYLEGIRK NFDEAAKVLK FNCEENQHSD SCYKLGAYYV TGKGGLTQDL KAAARCFLMA CEKPGKKSIA ACHNVGLLAH DGQVNEDGQP DLGKARDYYT RACDGGYTSS CFNLSAMFLQ GAPGFPKDMD LACKYSMKAC DLGHIWACAN ASRMYKLGDG VDKDEAKAEV LKNRAQQLHR EQQKGVQPLT FG (1-231 aa encoded by BC015313)
Required for assembly of mitochondrial respiratory chain complex I and complex IV.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-lactamase hcp-like protein; Cytochrome c oxidase assembly factor 7; hcp beta-lactamase-like protein C1orf163; hcp beta-lactamase-like protein C1orf163 homolog; respiratory chain assembly 1; Respiratory chain assembly factor 1; Sel1 repeat containing 1; Sel1 repeat-containing protein 1
Gene Aliases: 2010305A19Rik; AI451324; C1orf163; COA7; D4Ertd796e; RESA1; SELRC1
UniProt ID: (Human) Q96BR5, (Mouse) Q921H9
Entrez Gene ID: (Human) 65260, (Mouse) 69893
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support