Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The C1orf146 gene, also known as FAM213A, encodes a protein involved in antioxidative and anti-inflammatory processes within the cell. Located on chromosome 1, FAM213A (family with sequence similarity 213 member A) has been implicated in several critical cellular functions, including the regulation of reactive oxygen species (ROS) and modulation of inflammatory responses. The protein encoded by C1orf146 is known to interact with peroxiredoxins, which are crucial for protecting cells from oxidative damage. Research has shown that alterations in FAM213A expression can affect cellular susceptibility to oxidative stress and may play a role in the pathogenesis of various diseases, including cancer and inflammatory disorders.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein SPO16 homolog; RP11-163M2.4; Synaptonemal complex reinforcing element
Gene Aliases: C1orf146; SCRE; SPO16
UniProt ID: (Human) Q5VVC0
Entrez Gene ID: (Human) 388649
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support