Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDDDLMLALR LQEEWNLQEA ERDHAQESLS LVDASWELVD PTPDLQALFV QFNDQFFWGQ LEAVEVKWSV RMTLCAGICS YEGKGGMCSI RLSEPLLKLR PRKDLVETLL HEMIHAYLFV TNNDKDREGH GPEFCKHMHR INSLTGANIT VYHTFHDEVD EYRRHWWRCN GPCQHRPPYY GYVKRATNRE PSAHDYWWAE HQKTCGGTYI KIKEPENYSK KGKGKAKLGK EPVLAAENKD KPNRGEAQLV IPFSGKGYVL GETSNLPSPG KLITSHAINK TQDLLNQNHS ANAVRLNSKI KVKFEQNGSS KNSHLVSPAV SNSHQNVLSN YFPRVSFANQ KAFRGVNGSP RISVTVGNIP KNSVSSSSQR RVSSSKISLR NSSKVTESAS VMPSQDVSGS EDTFPNKRPR LEDKTVFDNF FIKKEQIKSS GNDPKYSTTT AQNSSSSSSQ SKMVNCPVCQ NEVLESQINE HLDWCLEGDS IKVKSEESL
SPRTN is a regulator of UV-induced DNA damage response: acts as a 'reader' of ubiquitinated PCNA that enhances RAD18-mediated PCNA ubiquitination and translesion DNA synthesis. SPRTN is recruited to sites of UV damage and interacts with ubiquitinated PCNA and RAD18, the E3 ubiquitin ligase that monoubiquitinates PCNA. It facilitates chromatin association of RAD18 and is required for efficient PCNA monoubiquitination, promoting a feed-forward loop to enhance PCNA ubiquitination and translesion DNA synthesis. It is also required for localization of DNA polymerase eta (POLH) to sites of UV damage.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DDDL1880; dJ876B10.3; DNA damage protein targeting VCP; DNA damage-targeting VCP (p97) adaptor; DNA-dependent metalloprotease SPRTN; DVC1; Protein with SprT-like domain at the N terminus; Spartan; zinc finger RAD18 domain-containing protein
Gene Aliases: C1orf124; DVC1; PRO4323; spartan; SPRTN; UNQ1880/PRO4323
UniProt ID: (Human) Q9H040
Entrez Gene ID: (Human) 83932
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support