FIGURE: 1 / 1
Immunogen sequence: LLLGTGKSPR QSLFFYPSYP DEVRGVFAVR TGKYKAHFFT QGSAHSDTTA DPACHASSSL TAHEPPLLYD LSKDPGENYN LLGGVAGATP EVLQALKQLQ LLKAQLDAAV TFGPSQVARG EDPALQICCH PGCTPRPACC HCP
The protein encoded by this gene hydrolyzes cerebroside sulfate to cerebroside and sulfate. Defects in this gene lead to metachromatic leucodystrophy, a progressive demyelination disease which results in a variety of neurological symptoms and ultimately death. Alternatively spliced transcript variants have been described for this gene.