Recommended positive controls: A431, H1299, HeLa, HepG2.
Predicted reactivity: Mouse (95%), Rat (95%), Xenopus laevis (86%), Rabbit (96%), Chicken (88%), Bovine (94%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the annexin family, a group of calcium-dependent phospholipid-binding proteins. Annexins have unique N-terminal domains and conserved C-terminal domains, which contain the calcium-dependent phospholipid-binding sites. The encoded protein is a 56-kD antigen recognized by sera from patients with various autoimmune diseases. Transcript variants encoding the same isoform have been identified.