Search Thermo Fisher Scientific
ELTI
The antibody may be used in ELISA and IAC. For ELISA, the recommended dilution is 1:200, depending on the conjugate used for detection. For positive controls ELISA plates are coated with 400 ng/mL BSA-conjugated C6/C8-alkylphenol.
Alkylphenols are used as starting material for the production of surface active substances, detergents, pesticides, paints, textile and paper. It is released during production and use of these products. Alkylphenols are recognized as endocrine disruptors, in particular estrogenic endocrine disruptors. A highly comparable compound is octylphenol.