Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ATP2C1 Antibodies

          Abnova

          ATP2C1 Polyclonal Antibody, MaxPab™

          View all (12) ATP2C1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ATP2C1 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (3)
          ATP2C1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ATP2C1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ATP2C1 Antibody (H00027032-D01P) in WB

          Western Blot analysis of ATP2C1 expression in transfected 293T cell line (H00027032-T02) by ATP2C1 MaxPab polyclonal antibody.

          Lane 1: ATP2C1 transfected lysate(100.60 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ATP2C1 Antibody in Western Blot (WB)
          ATP2C1 Antibody in Western Blot (WB)
          ATP2C1 Antibody in Western Blot (WB)

          Product Details

          H00027032-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ATP2C1 (NP_055197.2, 1 a.a. approximately 919 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MKVARFQKIP NGENETMIPV LTSKKASELP VSEVASILQA DLQNGLNKCE VSHRRAFHGW NEFDISEDEP LWKKYISQFK NPLIMLLLAS AVISVLMHQF DDAVSITVAI LIVVTVAFVQ EYRSEKSLEE LSKLVPPECH CVREGKLEHT LARDLVPGDT VCLSVGDRVP ADLRLFEAVD LSIDESSLTG ETTPCSKVTA PQPAATNGDL ASRSNIAFMG TLVRCGKAKG VVIGTGENSE FGEVFKMMQA EEAPKTPLQK SMDLLGKQLS FYSFGIIGII MLVGWLLGKD ILEMFTISVS LAVAAIPEGL PIVVTVTLAL GVMRMVKKRA IVKKLPIVET LGCCNVICSD KTGTLTKNEM TVTHIFTSDG LHAEVTGVGY NQFGEVIVDG DVVHGFYNPA VSRIVEAGCV CNDAVIRNNT LMGKPTEGAL IALAMKMGLD GLQQDYIRKA EYPFSSEQKW MAVKCVHRTQ QDRPEICFMK GAYEQVIKYC TTYQSKGQTL TLTQQQRDVY QQEKARMGSA GLRVLALASG PELGQLTFLG LVGIIDPPRT GVKEAVTTLI ASGVSIKMIT GDSQETAVAI ASRLGLYSKT SQSVSGEEID AMDVQQLSQI VPKVAVFYRA SPRHKMKIIK SLQKNGSVVA MTGDGVNDAV ALKAADIGVA MGQTGTDVCK EAADMILVDD DFQTIMSAIE EGKGIYNNIK NFVRFQLSTS IAALTLISLA TLMNFPNPLN AMQILWINII MDGPPAQSLG VEPVDKDVIR KPPRNWKDSI LTKNLILKIL VSSIIIVCGT LFVFWRELRD NVITPRDTTM TFTCFVFFDM FNALSSRSQT KSVFEIGLCS NRMFCYAVLG SIMGQLLVIY FPPLQKVFQT ESLSILDLLF LLGLTSSVCI VAEIIKKVER SREKIQKHVS STSSSFLEV

          Target Information

          ATP2C1, also known as secretory pathway Ca2+/Mn2+-ATPase (SPCA) 1, belongs to the family of P-type cation transport ATPases. This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the transport of the calcium from the cytosol to the Golgi lumen. Defects in this gene cause Hailey-Hailey disease, an autosomal dominant disorder characterized by persistent blisters and erosions of the skin. Unlike the related protein ATP2C2, ATP2C1 is ubiquitously expressed and displays a lower maximal turnover rate for overall Ca2+-ATPase reaction and a higher apparent affinity for cytosolic Ca2+ activation of phosphorylation. Recent evidence suggests that ATP2C1 is a key regulator of insulin-like growth factor receptor (IGF1R) processing in tumor progression in basal breast cancers.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ATP-dependent Ca(2+) pump PMR1; ATPase 2C1; ATPase, Ca(2+)-sequestering; ATPase, Ca++ transporting, type 2C, member 1; Ca(2+)/Mn(2+)-ATPase 2C1; Calcium-transporting ATPase type 2C member 1; HUSSY 28; HUSSY-28; rat; Secretory pathway Ca(2+)-transporting ATPase type 1; secretory pathway Ca2+/Mn2+ ATPase 1; SPCA1

          View more View less

          Gene Aliases: 1700121J11Rik; ATP2C1; ATP2C1A; AW061228; BCPM; D930003G21Rik; HHD; hSPCA1; HUSSY-28; KIAA1347; PMR1; PMR1L; SPCA; SPCA1

          View more View less

          UniProt ID: (Human) P98194, (Mouse) Q80XR2

          View more View less

          Entrez Gene ID: (Human) 27032, (Mouse) 235574

          View more View less

          Function(s)
          signal transducer activity calcium-transporting ATPase activity ATP binding metal ion binding nucleotide binding calcium ion binding manganese-transporting ATPase activity hydrolase activity manganese ion binding
          Process(es)
          calcium ion transport cellular calcium ion homeostasis signal transduction epidermis development calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules actin cytoskeleton reorganization Golgi calcium ion homeostasis Golgi calcium ion transport ion transmembrane transport positive regulation of I-kappaB kinase/NF-kappaB signaling calcium ion transmembrane transport transport ion transport manganese ion transport cellular manganese ion homeostasis manganese ion transmembrane transport
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0