Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MSWSPEECKG QGEPLDDRHP LCARLVEKPS RGSEEHLKSG PGPIVTRTAS GPALAFWQAV LAGDVGCVSR ILADSSTGLA PDSVFDTSDP ERWRDFRFNI RALRLW
May be a substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat and SOCS box protein 10; ankyrin repeat and SOCS box-containing protein 10; ASB-10
Gene Aliases: Asb-10; ASB10; BB137407; GLC1F; RGD1562922
UniProt ID: (Human) Q8WXI3, (Mouse) Q91ZT7
Entrez Gene ID: (Human) 136371, (Mouse) 117590, (Rat) 499972
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support