Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
May be involved in the cell-cell adhesion. May play a role in adipocyte differentiation and development of obesity. Is required for normal small intestine development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adipocyte adhesion molecule; adipocyte-specific adhesion molecule; Adipocyte-specific protein 5; CAR-like membrane protein; Coxsackie- and adenovirus receptor-like membrane protein; CXADR-like membrane protein; FLJ22415; visceral adipose tissue-specific transmembrane protein OL-16
Gene Aliases: 9030425E11Rik; ACAM; ASAM; ASP5; AW557819; CLMP; CSBM; CSBS; Ol16; UNQ318/PRO363
UniProt ID: (Human) Q9H6B4, (Mouse) Q8R373, (Rat) Q8K1G0
Entrez Gene ID: (Human) 79827, (Mouse) 71566, (Rat) 286939
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support