Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: GLSDQTSIL SNLPSFQSFH IVILGLDCAG KTTVLYRLQF NEFVNTVPTK GFNTEKIKVT LGNSKTVTFH FWDVGGQEKL RPLWKSYTRC TDGIVFVVDS VDVERMEEAK TELHKITRIS ENQGVPVLIV ANKQDLRNSL SLSEIEKLLA MGELSSSTPW HLQPTCAIIG DGL (4-175 aa encoded by BC001111)
ARL4A (ADP-ribosylation factor-like 4A) is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4A is similar to ARL4C and ARL4D and each has a nuclear localization signal and an unusually high guaninine nucleotide exchange rate. ARL4A is located in both the nuclear and extranuclear cell compartments. Multiple transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factor like GTPase 4A; ADP-ribosylation factor-like 4; ADP-ribosylation factor-like 4A; ADP-ribosylation factor-like protein 4A; ADP-ribosylation-like 4; OTTHUMP00000155200; OTTHUMP00000201575
Gene Aliases: AI467555; ARL4; ARL4A
UniProt ID: (Human) P40617, (Mouse) P61213, (Rat) P61214
Entrez Gene ID: (Human) 10124, (Mouse) 11861, (Rat) 29308
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support