Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive test controls include: A-431, LO2, HeLa, Mouse eye, Mouse brain, Mouse heart, Mouse kidney, Rat brain. The target is usually found in the following locations: Cell projection, Cytoplasm, Cytoplasmic side, Golgi apparatus membrane, Nucleus, Peripheral membrane protein, centrosome, cilium, cytoskeleton, microtubule organizing center, spindle.
Immunogen sequence: MGLLSILRKL KSAPDQEVRI LLLGLDNAGK TTLLKQLASE DISHITPTQG FNIKSVQSQG FKLNVWDIGG QRKIRPYWKN YFENTDILIY VIDSADRKRF EETGQELAEL LEEEKLSCVP VLIFANKQDL LTAAPASEIA EGLNLHTIRD RVWQIQSCSA LTGEGVQDGM NWVCKNVNAK KK
ADP-ribosylation factors (ARFs) are low molecular weight GTP-binding proteins belonging to the RAS superfamily. The predicted 182-amino acid ARL3 (ADP-ribosylation like factor) protein shares 97% amino acid identity with rat ARLl3 and 43% identity with human ARF1. Like the ARFs, ARL3 has a glycine at position 2, the site of N myristoylation, and lacks cysteine residues near the C terminus, which are found in other members of the RAS family. Northern blot analysis detected a 1-kb ARL3 transcript in all tissues tested, with highest expression in heart and lung, and lower expression in brain, liver, kidney, ovary, and testis. A 5.5-kb transcript was also detected in most tissues, with highest expression in brain. Immunoblot analysis detected ARL3 in human tumor cell lines but not in normal rodent cells. Although ARL3 binds GTP, it is devoid of activity in the cholera toxin-dependent ADP-ribosylation of Gs, and is therefore classified as an ARF-like protein.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factor like GTPase 3; ADP-ribosylation factor-like 3; ADP-ribosylation factor-like protein 3; ADP-ribosylation-like 3; ARD3; ARF-like 3; Arf-like protein 3
Gene Aliases: ARFL3; ARL3
UniProt ID: (Human) P36405, (Rat) P37996, (Mouse) Q9WUL7
Entrez Gene ID: (Human) 403, (Rat) 64664, (Mouse) 56350
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support