Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: TLDTDDRSE QEGSTLSVIS ATSSAGRLLP PRERLREVAF EYCQRLIEQS NRRALRKGDS DLQKACLVEA VLVLDVLCRQ DPSFLYRSLS CLKALHGRVR GDPASV (260-364 aa encoded by BC037399)
The function of this protein has not been specifically defined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adapter-related protein complex 5 zeta subunit; adaptor related protein complex 5, zeta 1 subunit; Adaptor-related protein complex 5 zeta subunit; adaptor-related protein complex 5, zeta 1 subunit; AP-5 complex subunit zeta-1; protein KIAA0415; Zeta5
Gene Aliases: AP5Z1; KIAA0415; SPG48; zeta
UniProt ID: (Human) O43299
Entrez Gene ID: (Human) 9907
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support