Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MLLLDCNPEV DGLKHLLETG ASVNAPPDPC KQSPVHLAAG SGLACFLLWQ LQTGADLNQQ DVLGEAPLHK AAKVGSLECL SLLVASDAQI DLCNKNGQTA EDLAWSCGFP DCAKFLTTIK CMQTIKASEH PDRNDCVAVL RQKRSLGSVE NTSGKRKC
ANKRD37 (Ankyrin Repeat Domain 37) is a Protein Coding gene. An important paralog of this gene isANKRD42.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat domain-containing protein 37; hLrp2bp; low density lipoprotein receptor-related protein binding protein; Low-density lipoprotein receptor-related protein 2-binding protein
Gene Aliases: ANKRD37; LPR2BP; Lrp2bp
UniProt ID: (Human) Q7Z713
Entrez Gene ID: (Human) 353322
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support