Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MCRMSFKKET PLANAAFWAA RRGNLALLKL LLNSGRVDVD CRDSHGTTLL MVAAYAGHID CVRELVLQGA DINLQRESGT TALFFAAQQG HNDVVRFLFG FGASTEFRTK DEGTALLAAS QYGHMQVVET LLKHGANIHD QLYDGATALF LAAQGGYLDV IRLLLASGAK VNQPRQDGTA PLWIASQMGH SEVVRVMLLR GADRDAARND GTTALLKAAN KGYNDVIKEL LKFSPTLGIL KNGTSALHAA VLSGNIKTVA LLLEAGADPS LRNKANELPA ELTKNERILR LLRSKEGPRK S
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat domain-containing protein 29
Gene Aliases: ANKRD29
UniProt ID: (Human) Q8N6D5
Entrez Gene ID: (Human) 147463
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support