Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ALAS1 Antibodies

          OriGene

          ALAS1 Monoclonal Antibody (OTI5D5), TrueMAB™

          View all (17) ALAS1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ALAS1 Monoclonal Antibody (OTI5D5), TrueMAB™

          • Antibody Testing Data (1)
          ALAS1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ALAS1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ALAS1 Antibody (CF808194) in WB

          HEK293T cells were transfected with the pCMV6-ENTRY control (Left lane) or pCMV6-ENTRY ALAS1 (RC219938, Right lane) cDNA for 48 hrs and lysed. Equivalent amounts of cell lysates (5 µg per lane) were separated by SDS-PAGE and immunoblotted with anti-ALAS1. Positive lysates LY400229 (100 µg) and LC400229 (20 µg) can be purchased separately from OriGene. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ALAS1 Antibody in Western Blot (WB)

          Product Details

          CF808194

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          OTI5D5

          Immunogen

          Human recombinant protein fragment corresponding to amino acids 57-308 of human ALAS1 produced in E.coli.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          1 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 8% trehalose

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

          Target Information

          ALAS-1 is a nuclear-encoded mitochondrial matrix enzyme which catalyzes the condensation of glycine with succinyl-CoA to form delta-amino levulinate, CO2 and CoA. It regulates the first and rate-limiting step of heme biosynthetic pathway. It is one of the two isoforms of ALAS and is a pyridoxal-5' phosphate dependant housekeeping enzyme. It is ubiquitously expressed and is responsible of providing heme for cytochromes and other hemoproteins. ALAS1 in liver undergoes negative feedback regulation by heme. Transcription of ALAS1 gene is stimulated by cAMP and respiratory uncoupling whereas Phorbol esters and insulin repress ALAS1 gene expression. ALAS1 is suggested to be involved in reciprocal regulation of Heme biosynthesis and circadian clock suggesting a potential target for treatment of circadian disorders. ALAS1 gene expression is down regulated in Acute Liver failure resulting in altered heme metabolism and liver function.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 5-aminolevulinate synthase 1; 5-aminolevulinate synthase, non-specific, mitochondrial; 5-aminolevulinate synthase, nonspecific, mitochondrial; 5-aminolevulinic acid synthase 1; ALA-S; ALAS-H; ALAS-N; aminolevulinate synthase H; aminolevulinate, delta-, synthase 1; aminolevulinic acid synthase 1; Delta-ALA synthase 1; delta-ALA synthetase; Delta-aminolevulinate synthase 1; migration-inducing protein 4; succinyl-CoA: glycine C-succinyl transferase

          View more View less

          Gene Aliases: ALAS; Alas-1; ALAS-H; ALAS-N; ALAS1; ALAS3; ALASH; MIG4; OK/SW-cl.121

          View more View less

          UniProt ID: (Human) P13196, (Rat) P13195, (Mouse) Q8VC19

          View more View less

          Entrez Gene ID: (Human) 211, (Rat) 65155, (Mouse) 11655

          View more View less

          Function(s)
          5-aminolevulinate synthase activity protein binding pyridoxal phosphate binding catalytic activity transferase activity transferase activity, transferring acyl groups
          Process(es)
          response to hypoxia protoporphyrinogen IX biosynthetic process response to herbicide response to organic substance response to nickel cation response to organic cyclic compound response to nutrient levels response to cobalt ion cellular response to insulin stimulus response to gonadotropin response to drug response to ethanol response to cAMP response to platinum ion cellular response to organic cyclic compound porphyrin-containing compound metabolic process heme biosynthetic process metabolic process biosynthetic process tetrapyrrole biosynthetic process mitochondrion organization
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-588f58b695-w7k2p:80/100.66.134.75:80.
          git-commit: b8205085541c8e5054bc4cc0f27ee9f88e66ec64
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0