FIGURE: 1 / 1
Predicted to react with all mammals based on sequence homology.
Heat-mediated antigen retrieval is recommended for the staining of paraffin sections. A suggested positive control for immunohistochemical applications is normal brain tissue. In Western blot, this antibody detects a band at ~30kDa in HEK293 cell lysate.
14-3-3 gamma encodes a product that belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 100% identical to the rat ortholog. It is induced by growth factors in human vascular smooth muscle cells, and is also highly expressed in skeletal and heart muscles, suggesting an important role for this protein in muscle tissue. It has been shown to interact with RAF1 and protein kinase C, proteins involved in various signal transduction pathways.