Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MAPLKGVQEQ QVRHECEVTK PEKEKGEETR IENGKLIVVT DSCGRVESSG KISEPMEAHN EGSNLESHQA KPKEKIEYKC SEREQRFIQH LDLIEHASTH TGKKLCESDV CQSSSLTGHK KVLSREKGHQ CHECGKAFQR SSHLVRHQKI HLGEKPYQCN ECGKVFSQNA GLLEHLRIHT GEKPYLCIHC GKNFRRSSHL NRHQRIHSQE EPCECKECGK TFSQALLLTH HQRIHSHSKS HQCNECGKAF SLTSDLIRHH RIHTGEKPFK CNICQKAFRL NSHLAQHVRI HNEEKPYQCS ECGEAFRQRS GLFQHQRYHH KDKLA
ZNF187 may be involved in transcriptional regulation (Probable).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein SRE-ZBP; Zinc finger and SCAN domain-containing protein 26; Zinc finger protein 187
Gene Aliases: SRE-ZBP; SREZBP; ZNF187; ZSCAN26
UniProt ID: (Human) Q16670
Entrez Gene ID: (Human) 7741
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support