PA1-31524 detects Thromboxane B2 from bovine samples.
PA1-31524 has been successfully used in radioimmune assay applications.
The PA1-31524 immunogen is: Highly pure Thromboxane B2 conjugated to bovine albumin
Thromboxane B2 is an inactive metabolite/product of thromboxane A2. It itself is not involved in platelet activation and aggregation in case of a wound, but its precursor, thromboxane A2 is. Thromboxane A2 synthesis is the target of the drug aspirin, which inhibits the COX-1 enzyme (the source of thromboxane A2 in platelets).