Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MSRMFCQAAR VDLTLDPDTA HPALMLSPDR RGVRLAERRQ EVADHPKRFS ADCCVLGAQG FRSGRHYWEV EVGGRRGWAV GAARESTHHK EKVGPGGSSV GSGDASSSRH HHRRRRLHLP QQPLLQREVW CVGTNGKRYQ AQSSTEQTLL SPSEKPRRFG VYLDYEAGRL GFYNAETLAH VHTFSAAFLG ERVFPFFRVL SKGTRIKLCP
TRIM41 belongs to a family of tripartite motif (TRIM) proteins defined as containing a RING finger, one or more B-box domains, and a coiled-coil region (Tanaka et al., 2005 ). See TRIM45.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRIM41; RINCK; RING finger-interacting protein with C kinase; RING-finger protein that interacts with C kinase; tripartite motif protein 41; Tripartite motif-containing protein 41
Gene Aliases: AW552703; BC020156; R75223; RINCK; TRIM41
UniProt ID: (Human) Q8WV44, (Mouse) Q5NCC3
Entrez Gene ID: (Human) 90933, (Mouse) 211007, (Rat) 303088
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support