Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: VEETRYCVD QNLLKEYFPV QVVTHGLLGI YQELLGLAFH HEEGASAWHE DVRLYTARDA ASGEVVGKFY LDLYPREGKY GHAACFGLQP GCLRQDGSRQ IAIAAMVANF TKPTADAPSL LQHDEVETYF HEFGHVMHQL CSQAEFAMFS GTHVERDFVE APSQMLENWV WEQEPLLRMS RHYRTGSAVP RELLEKLIES RQANTGLFNL RQIVLAKVDQ ALHTQTDADP AEEYARLCQE ILGVPATPGT NMPATFGHLA GGYDAQYYGY LWSEVYSMDM FHTRFKQEGV LNSKVGMDYR SCILRPGGSE DASAMLRRFL GRDPKQDAFL LSKGLQVGGC EPEPQVC (344-689 aa encoded by BC000135 )
A steady-state level of amyloid beta-peptide (Abeta) represents a balance between its biosynthesis from the amyloid precursor protein (APP) through the action of the beta and gamma-secretases and its catabolism by a variety of proteolytic enzymes. Thimet Oligopeptidase (THOP1) has been shown to promote A-beta degradation. It has also been shown to play a role in antigen presentation, metabolism of neuropeptides under 20 amino acids, and cytoplasmic peptide degradation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 3.4.24; EC 3.4.24.15; Endo-oligopeptidase A; Endopeptidase 24.15; metalloendopeptidase; MP78; PZ-peptidase; Soluble metallo-endopeptidase; Thimet oligopeptidase
Gene Aliases: AI131655; AI327041; EP24.15; MEPD_HUMAN; MP78; THOP1; TOP
UniProt ID: (Human) P52888, (Mouse) Q8C1A5, (Rat) P24155
Entrez Gene ID: (Human) 7064, (Mouse) 50492, (Rat) 64517
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support