Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: EACLEPQITP SYYTTSDAVI STETVLIVEI SLTCKNRVQN MALYADVGGK QFPVTRGQDV GRYQVSWSLD HKSAHAGTYE VRFFDEESYS LLRKAQRNNE DISIIPPLFT VSVDHRGTWN GPWVSTEVLA AAIGLVIYYL AFSAKSHIQA
TRAP proteins are part of a complex whose function is to bind calcium to the ER membrane and thereby regulate the retention of ER resident proteins. SSR4. also called TRAPD. is assumed to be involved in protein secretion. It is located in the Xq28 region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Signal sequence receptor subunit delta; signal sequence receptor, delta; SSR-delta; Translocon-associated protein subunit delta; TRAP-delta
Gene Aliases: CDG1Y; SSR4; TRAPD
UniProt ID: (Human) P51571
Entrez Gene ID: (Human) 6748
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support