Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MDKQSSAGG VKRSVPCESN EVNETMPETP TGDSDPQPAP KKMKTSESST ILVVRYRRNV KRTSPEELLN DHARENRINP LQMEEEEFME IMVEIPAK (1-97 aa encoded by BC054023)
Temporally regulated transcription and translation of several testis-specific genes is required to initiate the series of molecular and morphological changes in the male germ cell lineage necessary for the formation of mature spermatozoa. This gene is a member of the SPANX family, which is located in a gene cluster on chromosome X. The SPANX genes encode differentially expressed testis-specific proteins that localize to various subcellular compartments. This particular gene encodes a protein that localizes to the nucleus and is expressed in highly metastatic cell lines, making the protein a potential diagnostic and prognostic marker. The protein belongs to a family of cancer/testis antigens and represents a potential target for cancer immunotherapy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer-testis-associated protein CTp11; Cancer/testis antigen 11.3; cancer/testis antigen family 11, member 3; Cancer/testis-associated protein CTp11; cancer/testis-associated protein of 11 kD; CT11.3; Nuclear-associated protein SPAN-Xc; SPAN-Xc protein; SPANX family member C; SPANX family, member C; SPANX-C; Sperm protein associated with the nucleus on the X chromosome C; sperm protein associated with the nucleus, X chromosome, family member C
Gene Aliases: C; CT11.3; CTp11; SPANX-C; SPANXC
UniProt ID: (Human) Q9NY87
Entrez Gene ID: (Human) 64663
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support