Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MTSANKAIE LQLQVKQNAE ELQDFMRDLE NWEKDIKQKD MELRRQNGVP EENLPPIRNG NFRKKKKGKA KESSKKTREE NTKNRIKSYD YEAWAKLDVD RILDELDKDD STHESLSQES ESEEDGIHVD SQKALVLKEK GNKYFKQGKY DEAIDCYTKG MDADPYNPVL PTNRASAYFR LKKFAVAESD CNLAVALNRS YTKAYSRRGA ARFALQKLEE AKKDYERVLE LEPNNFEATN ELRKISQALA SKENSYPKEA DIVIKSTEGE RKQIEAQQNK QQAISEKDRG NGFFKEGKYE RAIECYTRGI AADGANALLP ANRAMAYLKI QKYEEAEKDC TQAILLDGSY SKAFARRGTA R (1-360 aa encoded by BC056415)
This gene encodes an RNA polymerase II-associated protein. The encoded protein may function in transcriptional regulation and may also regulate apoptosis. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RNA polymerase II-associated protein 3
Gene Aliases: RPAP3
UniProt ID: (Human) Q9H6T3
Entrez Gene ID: (Human) 79657
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support