Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • mTOR Antibodies

          Invitrogen

          Phospho-mTOR (Ser2481) Recombinant Rabbit Monoclonal Antibody (9Y5P6)

          View all (97) mTOR antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-mTOR (Ser2481) Recombinant Rabbit Monoclonal Antibody (9Y5P6)

          • Antibody Testing Data (1)
          Phospho-mTOR (Ser2481) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-mTOR (Ser2481) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-mTOR (Ser2481) Antibody (MA5-35838) in WB

          Western blot analysis of Phospho-mTOR (Ser2481) in extracts of various cell lines. Samples were incubated with Phospho-mTOR (Ser2481) Monoclonal antibody (Product # MA5-35838) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) secondary antibody at a dilution of 1:10,000. Both 293T cells and NIH/3T3 cells were treated by IGF-1 (50 ng/mL) at 37°C for 5 minutes after serum-starvation over... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-mTOR (Ser2481) Antibody in Western Blot (WB)

          Product Details

          MA5-35838

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          9Y5P6

          Immunogen

          A synthetic phosphorylated peptide around S2481 of human mTOR (P42345)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.5 mg/mL

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.05% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2849737

          Product Specific Information

          Immunogen sequence: IHSFI

          Target Information

          FRAP1 (mTOR) is a serine/threonine kinase that plays a critical role in cellular growth and proliferation. Perturbations in the mTOR/PI3-kinase/AKT pathway are associated with numerous forms of cancer. FRAP1 is also the target of rapamycin and its analogues, which are currently used as immunosuppressants and cancer therapeutics. Mutations affecting the gene results in Smith-Kingsmore syndrome.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: angiopoietin-like factor CDT6; FK506 binding protein 12-rapamycin associated protein 1; FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP-rapamycin associated protein (FRAP); FKBP-rapamycin-associated protein FRAP; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; m-TOR; Mammalian target of rapamycin; Mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); mTOR; mTORC1; Rapamycin and FKBP12 target 1; rapamycin associated protein FRAP2; Rapamycin target protein 1; RAPT1; Serine/threonine-protein kinase mTOR; Tyrosine-protein kinase mTOR

          View more View less

          Gene Aliases: 2610315D21Rik; AI327068; flat; FRAP; FRAP1; FRAP2; MTOR; RAFT1; RAPT1; SKS

          View more View less

          UniProt ID: (Human) P42345, (Mouse) Q9JLN9

          View more View less

          Entrez Gene ID: (Human) 2475, (Mouse) 56717

          View more View less

          Function(s)
          RNA polymerase III type 1 promoter DNA binding RNA polymerase III type 2 promoter DNA binding RNA polymerase III type 3 promoter DNA binding TFIIIC-class transcription factor binding protein kinase activity protein serine/threonine kinase activity protein binding ATP binding kinase activity protein kinase binding protein domain specific binding ribosome binding phosphoprotein binding nucleotide binding drug binding transferase activity transferase activity, transferring phosphorus-containing groups phosphotransferase activity, alcohol group as acceptor protein dimerization activity
          Process(es)
          negative regulation of protein phosphorylation positive regulation of protein phosphorylation positive regulation of endothelial cell proliferation heart morphogenesis heart valve morphogenesis regulation of glycogen biosynthetic process energy reserve metabolic process 'de novo' pyrimidine nucleobase biosynthetic process DNA repair protein phosphorylation response to stress cell cycle arrest signal transduction germ cell development brain development cell aging response to nutrient long-term memory visual learning post-embryonic development negative regulation of autophagy positive regulation of lamellipodium assembly positive regulation of gene expression positive regulation of myotube differentiation positive regulation of neuron maturation negative regulation of muscle atrophy cell growth macroautophagy negative regulation of macroautophagy phosphorylation peptidyl-serine phosphorylation peptidyl-threonine phosphorylation spinal cord development protein catabolic process positive regulation of actin filament polymerization T cell costimulation negative regulation of protein ubiquitination ruffle organization regulation of myelination cellular response to nutrient levels TOR signaling regulation of fatty acid beta-oxidation regulation of response to food response to insulin regulation of actin cytoskeleton organization social behavior multicellular organism growth growth wound healing response to cocaine regulation of GTPase activity response to amino acid response to morphine regulation of carbohydrate utilization positive regulation of nitric oxide biosynthetic process regulation of osteoclast differentiation positive regulation of translation negative regulation of cell size regulation of protein kinase activity positive regulation of transcription from RNA polymerase III promoter protein autophosphorylation positive regulation of lipid biosynthetic process phosphatidylinositol-mediated signaling mRNA stabilization positive regulation of smooth muscle cell proliferation positive regulation of oligodendrocyte differentiation positive regulation of peptidyl-tyrosine phosphorylation voluntary musculoskeletal movement positive regulation of stress fiber assembly negative regulation of NFAT protein import into nucleus positive regulation of protein kinase B signaling cardiac muscle cell development cardiac muscle contraction maternal process involved in female pregnancy positive regulation of glial cell proliferation positive regulation of dendritic spine development positive regulation of cell growth involved in cardiac muscle cell development cellular response to hypoxia regulation of brown fat cell differentiation regulation of membrane permeability regulation of cellular response to heat positive regulation of neuron death positive regulation of transcription of nuclear large rRNA transcript from RNA polymerase I promoter positive regulation of eating behavior positive regulation of cholangiocyte proliferation positive regulation of sensory perception of pain negative regulation of cholangiocyte apoptotic process positive regulation of granulosa cell proliferation positive regulation of skeletal muscle hypertrophy negative regulation of iodide transmembrane transport double-strand break repair via homologous recombination regulation of carbohydrate metabolic process cell projection organization
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0