Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • EphB1/EphB2 Antibodies

          Invitrogen

          Phospho-EphB1/EphB2 (Tyr778, Tyr780) Polyclonal Antibody

          View all (6) EphB1/EphB2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-EphB1/EphB2 (Tyr778, Tyr780) Polyclonal Antibody

          • Antibody Testing Data (1)
          Phospho-EphB1/EphB2 (Tyr778, Tyr780) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-EphB1/EphB2 (Tyr778, Tyr780) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-EphB1/EphB2 (Tyr778, Tyr780) Antibody (PA5-106133) in WB

          Western blot analysis of Phospho-EphB1/EphB2 (Tyr778, Tyr780) in serum treated K562 whole cell lysate (absence (-) or presence (+) of non-phospho and phospho peptide). Samples were incubated with Phospho-EphB1/EphB2 (Tyr778, Tyr780) polyclonal antibody (Product # PA5-106133). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-EphB1/EphB2 (Tyr778, Tyr780) Antibody in Western Blot (WB)

          Product Details

          PA5-106133

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:3,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human EPHB1(Accession P54762), corresponding to amino acid residues around phosphorylated Tyr778.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          sequential chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2817531

          Product Specific Information

          Antibody detects endogenous levels of EPHB1/2 only when phosphorylated at Tyr778/Tyr780.

          Target Information

          Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase family. The protein encoded by this gene is a receptor for ephrin-B family members.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Developmentally-regulated Eph-related tyrosine kinase; EK5; EK6; ELK; ELK-related protein tyrosine kinase; ELK-related tyrosine kinase; Eph receptor B2 (ELK-related protein tyrosine kinase); EPH tyrosine kinase 2; EPH tyrosine kinase 3; EPH-like kinase 5; EPH-like kinase 6; EPHB3; Ephrin B1; Ephrin B2; Ephrin type-B receptor 1; Ephrin type-B receptor 2; EphrinB1; EphrinB2; ETK2; FLJ37986; HEK-6; HEK2; MGC87492; NET; Neural kinase; Neuronally-expressed EPH-related tyrosine kinase; Nuk receptor tyrosine kinase; protein-tyrosine kinase HEK5; Renal carcinoma antigen NY-REN-47; soluble EPHB1 variant 1; TYRO6; Tyrosine-protein kinase receptor EPH-2; Tyrosine-protein kinase receptor EPH-3; Tyrosine-protein kinase receptor SEK-3; Tyrosine-protein kinase TYRO5

          View more View less

          Gene Aliases: 9330129L11; AW488255; C130099E04Rik; CAPB; Cek5; Cek6; DRT; EK5; ELK; Elkh; ENSMUSG00000074119; EPHB1; EPHB2; EPHT2; EPHT3; Epth2; EPTH3; ERK; ETECK; HEK5; HEK6; NET; Nuk; PCBC; Prkm5; Qek5; RGD1564232; Sek3; TYRO5

          View more View less

          UniProt ID: (Human) P54762, (Human) P29323, (Rat) P09759, (Mouse) Q8CBF3, (Mouse) P54763

          View more View less

          Entrez Gene ID: (Human) 2047, (Human) 2048, (Rat) 24338, (Mouse) 270190, (Mouse) 13844, (Rat) 313633

          View more View less

          Function(s)
          transmembrane-ephrin receptor activity protein binding ATP binding axon guidance receptor activity protein complex binding protein tyrosine kinase activity receptor binding identical protein binding ephrin receptor activity nucleotide binding protein kinase activity transmembrane receptor protein tyrosine kinase activity kinase activity transferase activity transferase activity, transferring phosphorus-containing groups receptor activity molecular_function
          Process(es)
          angiogenesis immunological synapse formation axon guidance peptidyl-tyrosine phosphorylation cranial nerve development optic nerve morphogenesis central nervous system projection neuron axonogenesis neurogenesis establishment of cell polarity retinal ganglion cell axon guidance cell-substrate adhesion regulation of JNK cascade protein autophosphorylation ephrin receptor signaling pathway camera-type eye morphogenesis detection of temperature stimulus involved in sensory perception of pain positive regulation of synapse assembly cell chemotaxis dendritic spine development dendritic spine morphogenesis neural precursor cell proliferation regulation of ERK1 and ERK2 cascade regulation of neuron death protein phosphorylation cell adhesion transmembrane receptor protein tyrosine kinase signaling pathway nervous system development phosphorylation urogenital system development axonal fasciculation learning corpus callosum development inner ear morphogenesis positive regulation of long-term neuronal synaptic plasticity negative regulation of axonogenesis regulation of body fluid levels palate development commissural neuron axon guidance cell morphogenesis multicellular organism development organ morphogenesis regulation of neuronal synaptic plasticity regulation of axonogenesis biological_process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0