Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MQLTRCCFVF LVQGSLYLVI CGQDDGPPGS EDPERDDHEG QPRPRVPRKR GHISPKSRPM ANSTLLGLLA PPGEAWGILG QPPNRPNHSP PPSAKVKKIF GWGDFYSNIK TVALNLLVTG KIVDHGNGTF SVHFQHNATG QGNISISLVP PSKAVEFHQE QQIFIEAKAS KIFNCRMGWE KVERGRRTSL CTHDPAKICS RDHAQSSATW SCSQPFKVVC VYIAFYSTDY RLVQKVCPDY NYHSDTPYYP SG
May be signaling molecules that resemble neuropeptides. Ligand for alpha-neurexins. Target protein is a member of a family of neuropeptide-like glycoproteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neurexophilin-3
Gene Aliases: KIAA1159; NPH3; NXPH3; UNQ687/PRO1327
UniProt ID: (Human) O95157
Entrez Gene ID: (Human) 11248
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support