Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MEFAELIKTP RVDNVVLHRP FYPAVEGTLC LTGHHLILSS RQDNTEELWL LHSNIDAIDK RFVGSLGTII IKCKDFRIIQ LDIPGMEECL NIASSIEALS TLDSITLMYP FFYRPMFEVI EDGWHSFLPE QEFELYSSAT SEWRLSYVNK EFAVCPSYPP IVTVPKSIDD EALRKVATFR HGGRFPVLSY YHKKNGMVIM RSGQPLTGTN GRRCKEDEKL INATLRAGKR GYIIDTRSLN VAQQTRAKGG; Positive Samples: HepG2, HeLa, Jurkat, A-549, Mouse brain, Rat brain, Rat lung; Cellular Location: Cytoplasm
This gene encodes a myotubularin-related protein that is atypical to most other members of the myotubularin-related protein family because it has no dual-specificity phosphatase domain. The encoded protein contains a double-helical motif similar to the SET interaction domain, which is thought to have a role in the control of cell proliferation. In mouse, a protein similar to the encoded protein binds with MTMR7, and together they dephosphorylate phosphatidylinositol 3-phosphate and inositol 1,3-bisphosphate.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP434K171; Inactive phosphatidylinositol 3-phosphatase 9; MGC126672; myotubularin related protein 8; Myotubularin-related protein 9
Gene Aliases: 9430075G12Rik; AA516943; AF073881; C8orf9; LIP-STYX; mMTMH3; Mtmr3; MTMR8; MTMR9
UniProt ID: (Human) Q96QG7, (Mouse) Q9Z2D0
Entrez Gene ID: (Human) 66036, (Mouse) 210376, (Rat) 282584
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support