Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: KGKDRTVAEG EECAREMGVD IIARSESLGP ISELYSDELS EPEHEAARPV YPPHEGREIP TPEPLKVTFK SQVKVEDDES LLL
Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 59%.
MURC induces RHOA activation and activates NPPA transcription and myofibrillar organization through the Rho/ROCK signaling pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Caveolae-associated protein 4; Muscle-related coiled-coil protein; Muscle-restricted coiled-coil protein
Gene Aliases: cavin-4; CAVIN4; MURC
UniProt ID: (Human) Q5BKX8
Entrez Gene ID: (Human) 347273
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support